Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35556869-35557372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
UniGene
Hs.472558
Gene Chromosome Location
Chr.20: 35542029-35557634 on Build GRCh38
Transcripts
NM_015966.2 NM_198398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5454831 c.998C>T p.S333L Chr.20: 35557076-35557076 on Build GRCh38
6656092 c.981C>T p.F327F Chr.20: 35557059-35557059 on Build GRCh38
3545766 c.1083C>T p.F361F Chr.20: 35557245-35557245 on Build GRCh38
3545764 c.961C>T p.Q321* Chr.20: 35557039-35557039 on Build GRCh38
5391928 c.955G>A p.G319S Chr.20: 35557033-35557033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143458154 Chr.20: 35557059-35557059 on Build GRCh38
rs375424267 Chr.20: 35557232-35557232 on Build GRCh38
rs201696699 Chr.20: 35557029-35557029 on Build GRCh38
rs200279726 Chr.20: 35557050-35557050 on Build GRCh38
rs994811695 Chr.20: 35557330-35557330 on Build GRCh38
rs749199060 Chr.20: 35556938-35556938 on Build GRCh38
rs748048612 Chr.20: 35556987-35556987 on Build GRCh38
rs148891536 Chr.20: 35557052-35557052 on Build GRCh38
rs928108260 Chr.20: 35557077-35557077 on Build GRCh38
rs541557488 Chr.20: 35557083-35557083 on Build GRCh38
rs750196406 Chr.20: 35557291-35557291 on Build GRCh38
rs574293663 Chr.20: 35556894-35556894 on Build GRCh38
rs753548653 Chr.20: 35557161-35557161 on Build GRCh38
rs770306026 Chr.20: 35557132-35557132 on Build GRCh38
rs757699656 Chr.20: 35556974-35556974 on Build GRCh38
rs552186319 Chr.20: 35557320-35557320 on Build GRCh38
rs772359227 Chr.20: 35557153-35557155 on Build GRCh38
rs763473232 Chr.20: 35557268-35557268 on Build GRCh38
rs921723179 Chr.20: 35557222-35557222 on Build GRCh38
rs755883023 Chr.20: 35557001-35557001 on Build GRCh38
rs760268080 Chr.20: 35557155-35557155 on Build GRCh38
rs756985234 Chr.20: 35557173-35557173 on Build GRCh38
rs752138106 Chr.20: 35557153-35557153 on Build GRCh38
rs8825 Chr.20: 35556975-35556975 on Build GRCh38
rs577375717 Chr.20: 35557035-35557035 on Build GRCh38
rs761050745 Chr.20: 35556952-35556952 on Build GRCh38
rs76270188 Chr.20: 35556978-35556978 on Build GRCh38
rs376350265 Chr.20: 35557097-35557097 on Build GRCh38
rs761193151 Chr.20: 35557210-35557211 on Build GRCh38
rs370433773 Chr.20: 35557137-35557137 on Build GRCh38
rs770219962 Chr.20: 35557265-35557265 on Build GRCh38
rs749178011 Chr.20: 35557007-35557007 on Build GRCh38
rs373409776 Chr.20: 35557036-35557036 on Build GRCh38
rs773930147 Chr.20: 35556984-35556985 on Build GRCh38
rs767690881 Chr.20: 35556945-35556945 on Build GRCh38
rs774948658 Chr.20: 35557272-35557272 on Build GRCh38
rs377061605 Chr.20: 35557128-35557128 on Build GRCh38
rs878995702 Chr.20: 35557335-35557335 on Build GRCh38
rs763561259 Chr.20: 35557140-35557140 on Build GRCh38
rs747170826 Chr.20: 35557119-35557119 on Build GRCh38
rs772070697 Chr.20: 35557103-35557103 on Build GRCh38
rs765811014 Chr.20: 35557060-35557060 on Build GRCh38
rs543824492 Chr.20: 35557002-35557002 on Build GRCh38
rs754577778 Chr.20: 35556981-35556981 on Build GRCh38
rs1014914370 Chr.20: 35557011-35557011 on Build GRCh38
rs370108311 Chr.20: 35557098-35557098 on Build GRCh38
rs372528560 Chr.20: 35557123-35557123 on Build GRCh38
rs761500442 Chr.20: 35557137-35557138 on Build GRCh38
rs769511767 Chr.20: 35556905-35556905 on Build GRCh38
rs192374012 Chr.20: 35556897-35556897 on Build GRCh38
rs753364175 Chr.20: 35557284-35557284 on Build GRCh38
rs774398430 Chr.20: 35556940-35556940 on Build GRCh38
rs1045225363 Chr.20: 35557024-35557024 on Build GRCh38
rs900215393 Chr.20: 35556951-35556951 on Build GRCh38
rs369685477 Chr.20: 35557033-35557033 on Build GRCh38
rs748419150 Chr.20: 35557264-35557264 on Build GRCh38
rs759658444 Chr.20: 35556944-35556944 on Build GRCh38
rs777526654 Chr.20: 35557090-35557090 on Build GRCh38
rs996035644 Chr.20: 35556962-35556962 on Build GRCh38
rs768814237 Chr.20: 35557236-35557236 on Build GRCh38
rs11554456 Chr.20: 35557022-35557022 on Build GRCh38
rs891459649 Chr.20: 35556934-35556934 on Build GRCh38
rs750284270 Chr.20: 35557179-35557179 on Build GRCh38
rs747827771 Chr.20: 35556993-35556993 on Build GRCh38
rs752335483 Chr.20: 35557081-35557081 on Build GRCh38
rs764379837 Chr.20: 35556961-35556961 on Build GRCh38
rs754492134 Chr.20: 35557071-35557071 on Build GRCh38
rs556224346 Chr.20: 35557266-35557266 on Build GRCh38
rs768280329 Chr.20: 35556932-35556933 on Build GRCh38
rs771530195 Chr.20: 35557270-35557270 on Build GRCh38
rs147972524 Chr.20: 35557224-35557224 on Build GRCh38
rs770599819 Chr.20: 35557212-35557212 on Build GRCh38
rs750999591 Chr.20: 35557067-35557067 on Build GRCh38
rs200258928 Chr.20: 35557175-35557175 on Build GRCh38
rs754304886 Chr.20: 35556970-35556970 on Build GRCh38
rs775717022 Chr.20: 35556950-35556950 on Build GRCh38
rs530404938 Chr.20: 35557307-35557307 on Build GRCh38
rs115747345 Chr.20: 35557289-35557289 on Build GRCh38
rs917593605 Chr.20: 35556986-35556986 on Build GRCh38
rs781224374 Chr.20: 35557247-35557247 on Build GRCh38
rs762044840 Chr.20: 35557129-35557129 on Build GRCh38
rs796259783 Chr.20: 35557147-35557147 on Build GRCh38
rs780103745 Chr.20: 35557104-35557104 on Build GRCh38
rs530329677 Chr.20: 35557100-35557100 on Build GRCh38
rs763610128 Chr.20: 35557160-35557160 on Build GRCh38
rs773585155 Chr.20: 35557184-35557186 on Build GRCh38
rs760052229 Chr.20: 35557277-35557277 on Build GRCh38
rs6060465 Chr.20: 35557042-35557042 on Build GRCh38
rs899054172 Chr.20: 35557321-35557321 on Build GRCh38
rs767063765 Chr.20: 35557144-35557144 on Build GRCh38
rs6060464 Chr.20: 35556902-35556902 on Build GRCh38
rs763670170 Chr.20: 35557279-35557279 on Build GRCh38
rs770979943 Chr.20: 35556939-35556939 on Build GRCh38

View Full Product Details