Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH4
Entrez Gene ID
54784
Gene Name
alkB homolog 4, lysine demethylase
Gene Aliases
ABH4
Chromosome Location
Chr.7: 102456220 - 102464874 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017621.3 NP_060091.1 2 - 3 359 99
XM_005250464.2 XP_005250521.1 2 - 3 252 99
GenBank mRNA AK000020.1 - 2 - 3 346 99
AK026097.1 - 2 - 3 257 99
AK222689.1 - 2 - 3 348 99
BC002820.2 - 2 - 3 326 99
BC017096.1 - 2 - 3 326 99
BX353099.2 - 2 - 3 360 99

View Full Product Details