Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIT1
Entrez Gene ID
54802
Gene Name
tRNA isopentenyltransferase 1
Gene Aliases
GRO1, IPPT, IPT, IPTase, MOD5, hGRO1
Chromosome Location
Chr.1: 39841031 - 39883511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312692.1 NP_001299621.1 6 - 7 779 108
NM_017646.5 NP_060116.2 8 - 9 1025 108
NR_132401.1 8 - 9 1021 108
NR_132402.1 7 - 8 879 108
NR_132403.1 7 - 8 875 108
NR_132404.1 7 - 8 875 108
NR_132405.1 7 - 8 871 108
NR_132406.1 6 - 7 762 108
NR_132407.1 5 - 6 639 108
NR_132408.1 5 - 6 635 108
NR_132409.1 4 - 5 496 108
NR_132410.1 4 - 5 522 108
NR_132412.1 3 - 4 383 108
NR_132415.1 8 - 9 1112 108
XM_005270954.2 XP_005271011.1 8 - 9 1261 108
XM_006710706.1 XP_006710769.1 8 - 9 1093 108
GenBank mRNA AF074918.1 - 8 - 9 1106 108
AK000068.1 - 6 - 7 642 108
AK074222.1 - 6 - 7 642 108
AK223478.1 - 8 - 9 1018 108
AK307910.1 - 8 - 9 109
AY303390.1 - 8 - 9 1019 108
AY702933.1 - 8 - 9 948 108
AY702934.1 - 8 - 9 944 108
AY702935.1 - 7 - 8 795 108
AY702936.1 - 7 - 8 802 108
AY702937.1 - 7 - 8 798 108
AY702938.1 - 7 - 8 798 108
AY702939.1 - 7 - 8 794 108
AY702940.1 - 6 - 7 685 108
AY702941.1 - 5 - 6 562 108
AY702942.1 - 5 - 6 558 108
AY702943.1 - 4 - 5 419 108
AY702944.1 - 4 - 5 445 108
AY702945.1 - 3 - 4 306 108
BC010741.1 - 6 - 7 741 108
BC128155.1 - 8 - 9 1019 108
BC128156.1 - 8 - 9 1106 108
CR457226.1 - 5 - 6 576 108

View Full Product Details