The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199076.2 | NP_951058.1 | 5 - 6 | 2331 | 75 |
GenBank mRNA | AK000071.1 | - | 5 - 6 | 800 | 75 |
AK023479.1 | - | 5 - 6 | 2292 | 75 |