The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005268469.2 | XP_005268526.1 | 7 - 8 | 1305 | 96 |
GenBank mRNA | AK000202.1 | - | 3 - 4 | 671 | 96 |
AK056861.1 | - | 5 - 6 | 1404 | 96 | |
AK122862.1 | - | 5 - 6 | 2740 | 96 |