The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017780.3 | NP_060250.2 | 12 - 13 | 3695 | 96 |
XM_011517553.2 | XP_011515855.1 | 12 - 13 | 3716 | 96 | |
XM_011517554.2 | XP_011515856.1 | 12 - 13 | 3694 | 96 | |
XM_011517555.2 | XP_011515857.1 | 12 - 13 | 3716 | 96 | |
XM_011517560.2 | XP_011515862.1 | 12 - 13 | 3716 | 96 | |
XM_017013612.1 | XP_016869101.1 | 12 - 13 | 3770 | 96 | |
XM_017013613.1 | XP_016869102.1 | 12 - 13 | 3716 | 96 | |
GenBank mRNA | AB037837.1 | - | 8 - 9 | 917 | 96 |
AK000368.1 | - | 5 - 6 | 649 | 96 | |
AK225354.1 | - | 5 - 6 | 649 | 96 |