The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243342.1 | NP_001230271.1 | 9 - 10 | 1475 | 58 |
NM_017950.3 | NP_060420.2 | 9 - 10 | 1475 | 58 | |
XM_005257492.4 | XP_005257549.1 | 9 - 10 | 1475 | 58 | |
XM_011524963.2 | XP_011523265.1 | 9 - 10 | 1512 | 58 | |
XM_011524964.2 | XP_011523266.1 | 3 - 4 | 444 | 58 | |
XM_011524965.2 | XP_011523267.1 | 9 - 10 | 1475 | 58 | |
XM_017024807.1 | XP_016880296.1 | 9 - 10 | 1475 | 58 | |
GenBank mRNA | AB046860.1 | - | 5 - 6 | 981 | 58 |
AK000760.1 | - | 9 - 10 | 1455 | 58 | |
BC035251.1 | - | 9 - 10 | 1468 | 58 |