Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC45A
Entrez Gene ID
55898
Gene Name
unc-45 myosin chaperone A
Gene Aliases
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
Chromosome Location
Chr.15: 90929980 - 90954093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039675.1 NP_001034764.1 7 - 8 1215 66
NM_001323619.1 NP_001310548.1 4 - 5 474 66
NM_001323620.1 NP_001310549.1 4 - 5 1012 66
NM_001323621.1 NP_001310550.1 4 - 5 819 66
NM_018671.4 NP_061141.2 4 - 5 864 66
XM_005254963.3 XP_005255020.1 4 - 5 584 66
XM_011521779.2 XP_011520081.1 7 - 8 1460 66
XM_011521780.2 XP_011520082.1 3 - 4 366 66
XM_017022407.1 XP_016877896.1 4 - 5 584 66
GenBank mRNA AB014729.1 - 4 - 5 406 66
AB014736.1 - 4 - 5 448 66
AK000905.1 - 3 - 4 742 66
AK125721.1 - 7 - 8 1215 66
AK291622.1 - 4 - 5 419 66
AK293538.1 - 4 - 5 588 66
AK296964.1 - 4 - 5 441 66
AK302979.1 - 4 - 5 481 66
BC006214.1 - 4 - 5 447 66
BC037992.1 - 4 - 5 447 66
BC045635.1 - 7 - 8 1215 66

View Full Product Details