Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 77286542-77287080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK5
Entrez Gene ID
26289
Gene Name
adenylate kinase 5
Gene Aliases
AK6
UniGene
Hs.559718
Gene Chromosome Location
Chr.1: 77281908-77559969 on Build GRCh38
Transcripts
NM_012093.3 NM_174858.2 XM_005270739.4 XM_006710572.3 XM_017001008.1 XM_017001009.1 XM_017001010.1 XM_017001011.1 XM_017001012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295786 c.192G>T p.K64N Chr.1: 77287072-77287072 on Build GRCh38
4009938 c.108A>G p.E36E Chr.1: 77286988-77286988 on Build GRCh38
6293101 c.178G>T p.V60L Chr.1: 77287058-77287058 on Build GRCh38
2242276 c.110A>G p.Y37C Chr.1: 77286990-77286990 on Build GRCh38
682063 c.126A>C p.L42F Chr.1: 77287006-77287006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044067121 Chr.1: 77286599-77286599 on Build GRCh38
rs938324210 Chr.1: 77286850-77286850 on Build GRCh38
rs1048375460 Chr.1: 77286890-77286890 on Build GRCh38
rs534860294 Chr.1: 77286680-77286680 on Build GRCh38
rs923170834 Chr.1: 77286787-77286787 on Build GRCh38
rs1035916157 Chr.1: 77286678-77286678 on Build GRCh38
rs769175141 Chr.1: 77286991-77286991 on Build GRCh38
rs138257308 Chr.1: 77286860-77286864 on Build GRCh38
rs576514395 Chr.1: 77286739-77286739 on Build GRCh38
rs949556994 Chr.1: 77286889-77286889 on Build GRCh38
rs112791287 Chr.1: 77286636-77286636 on Build GRCh38
rs745690169 Chr.1: 77286890-77286891 on Build GRCh38
rs868604391 Chr.1: 77287017-77287017 on Build GRCh38
rs745640653 Chr.1: 77286745-77286745 on Build GRCh38
rs952824158 Chr.1: 77286712-77286712 on Build GRCh38
rs905542631 Chr.1: 77286840-77286841 on Build GRCh38
rs548675287 Chr.1: 77286571-77286571 on Build GRCh38
rs369882574 Chr.1: 77286918-77286918 on Build GRCh38
rs116187907 Chr.1: 77286634-77286634 on Build GRCh38
rs767219581 Chr.1: 77286928-77286932 on Build GRCh38
rs896745030 Chr.1: 77286869-77286869 on Build GRCh38
rs756637660 Chr.1: 77286959-77286959 on Build GRCh38
rs987038989 Chr.1: 77286748-77286748 on Build GRCh38
rs779228577 Chr.1: 77287000-77287000 on Build GRCh38
rs955173043 Chr.1: 77286746-77286746 on Build GRCh38
rs774426567 Chr.1: 77286570-77286570 on Build GRCh38
rs1055279554 Chr.1: 77286626-77286626 on Build GRCh38
rs749710378 Chr.1: 77286988-77286988 on Build GRCh38
rs962586448 Chr.1: 77286727-77286727 on Build GRCh38
rs146814780 Chr.1: 77286873-77286873 on Build GRCh38
rs776574133 Chr.1: 77286907-77286907 on Build GRCh38
rs185013024 Chr.1: 77286627-77286627 on Build GRCh38
rs886512893 Chr.1: 77286910-77286910 on Build GRCh38
rs867071635 Chr.1: 77286971-77286971 on Build GRCh38
rs190253352 Chr.1: 77286807-77286807 on Build GRCh38
rs538982404 Chr.1: 77286681-77286681 on Build GRCh38
rs913625892 Chr.1: 77286759-77286759 on Build GRCh38
rs185653281 Chr.1: 77287046-77287046 on Build GRCh38
rs945130363 Chr.1: 77286779-77286779 on Build GRCh38
rs373457972 Chr.1: 77286923-77286923 on Build GRCh38
rs938241394 Chr.1: 77286842-77286842 on Build GRCh38
rs934522039 Chr.1: 77286794-77286794 on Build GRCh38
rs567223905 Chr.1: 77286605-77286605 on Build GRCh38
rs546331554 Chr.1: 77286612-77286612 on Build GRCh38
rs1028105978 Chr.1: 77286740-77286740 on Build GRCh38
rs947078302 Chr.1: 77286565-77286565 on Build GRCh38
rs1027100829 Chr.1: 77286683-77286683 on Build GRCh38
rs559723762 Chr.1: 77286893-77286893 on Build GRCh38
rs750935740 Chr.1: 77286955-77286955 on Build GRCh38
rs748334363 Chr.1: 77287022-77287022 on Build GRCh38
rs557283813 Chr.1: 77286737-77286737 on Build GRCh38
rs765208716 Chr.1: 77287011-77287011 on Build GRCh38
rs370854870 Chr.1: 77287049-77287049 on Build GRCh38
rs201841905 Chr.1: 77286859-77286861 on Build GRCh38
rs555850127 Chr.1: 77286822-77286822 on Build GRCh38
rs751232412 Chr.1: 77286932-77286932 on Build GRCh38
rs752549497 Chr.1: 77286919-77286919 on Build GRCh38
rs1016249817 Chr.1: 77286708-77286708 on Build GRCh38
rs377289573 Chr.1: 77286974-77286974 on Build GRCh38
rs77711026 Chr.1: 77286864-77286864 on Build GRCh38
rs924964147 Chr.1: 77286832-77286832 on Build GRCh38
rs1006987807 Chr.1: 77286973-77286973 on Build GRCh38
rs386632620 Chr.1: 77286634-77286636 on Build GRCh38
rs375338556 Chr.1: 77287054-77287054 on Build GRCh38
rs947161585 Chr.1: 77286821-77286821 on Build GRCh38
rs11162308 Chr.1: 77286937-77286937 on Build GRCh38
rs759400171 Chr.1: 77286912-77286912 on Build GRCh38
rs561450114 Chr.1: 77286943-77286943 on Build GRCh38

View Full Product Details