The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018228.2 | NP_060698.2 | 1 - 2 | 196 | 94 |
GenBank mRNA | AK001673.1 | - | 1 - 2 | 150 | 94 |
BC053325.1 | - | 1 - 2 | 179 | 94 | |
CX165172.1 | - | 1 - 2 | 196 | 94 |