Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN3B
Entrez Gene ID
55800
Gene Name
sodium voltage-gated channel beta subunit 3
Gene Aliases
ATFB16, BRGDA7, HSA243396, SCNB3
Chromosome Location
Chr.11: 123629187 - 123654607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040151.1 NP_001035241.1 4 - 5 851 101
NM_018400.3 NP_060870.1 3 - 4 1250 101
XM_011542897.1 XP_011541199.1 4 - 5 851 101
GenBank mRNA AB032984.1 - 4 - 5 491 101
AJ243396.2 - 3 - 4 819 101
AK314513.1 - 4 - 5 838 101
AL136589.1 - 3 - 4 1247 101
BC117282.1 - 3 - 4 537 101
BC126265.1 - 3 - 4 537 101

View Full Product Details