Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC45A
Entrez Gene ID
55898
Gene Name
unc-45 myosin chaperone A
Gene Aliases
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
Chromosome Location
Chr.15: 90929980 - 90954093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039675.1 NP_001034764.1 20 - 21 3095 62
NM_001323619.1 NP_001310548.1 17 - 18 2354 62
NM_001323620.1 NP_001310549.1 17 - 18 2892 62
NM_001323621.1 NP_001310550.1 17 - 18 2699 62
NM_018671.4 NP_061141.2 17 - 18 2744 62
XM_005254963.3 XP_005255020.1 17 - 18 2464 62
XM_011521779.2 XP_011520081.1 20 - 21 3340 62
XM_011521780.2 XP_011520082.1 16 - 17 2176 62
XM_017022407.1 XP_016877896.1 17 - 18 2394 62
GenBank mRNA AB014729.1 - 17 - 18 2286 62
AB014736.1 - 17 - 18 2328 62
AK125523.1 - 6 - 7 1734 62
AK125721.1 - 20 - 21 3095 62
AK291622.1 - 17 - 18 2299 62
AL357537.1 - 2 - 3 571 62
AL357538.1 - 1 - 2 40 62
BC006214.1 - 17 - 18 2327 62
BC010995.2 - 4 - 5 334 62
BC037992.1 - 17 - 18 2327 62
BC045635.1 - 20 - 21 3095 62

View Full Product Details