The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024627.1 | 1 - 2 | 834 | 94 | |
GenBank mRNA | AB039920.1 | - | 1 - 2 | 834 | 94 |
AJ404617.2 | - | 1 - 2 | 804 | 94 | |
BC098159.1 | - | 1 - 2 | 696 | 94 | |
BC098256.1 | - | 1 - 2 | 696 | 94 | |
BC098319.1 | - | 1 - 2 | 700 | 94 | |
BC098344.1 | - | 1 - 2 | 696 | 94 |