The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018944.2 | NP_061817.1 | 1 - 2 | 389 | 84 |
XM_017028400.1 | XP_016883889.1 | 1 - 2 | 480 | 84 | |
XM_017028401.1 | XP_016883890.1 | 1 - 2 | 480 | 84 | |
GenBank mRNA | AF231921.1 | - | 1 - 2 | 361 | 84 |
AF387845.1 | - | 1 - 2 | 359 | 84 | |
AK075281.1 | - | 1 - 2 | 361 | 84 | |
AK312073.1 | - | 1 - 2 | 480 | 84 | |
BC042917.1 | - | 1 - 2 | 365 | 84 | |
BU616573.1 | - | 1 - 2 | 392 | 84 | |
CB959520.1 | - | - | 438 | 84 |