Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- STAG3L3
- Entrez Gene ID
-
442578
- Gene Name
- stromal antigen 3-like 3 (pseudogene)
- Gene Aliases
-
STAG3L1, STAG3L2, STAG3L3P
- Chromosome Location
-
Chr.7: 72997295 - 73005943 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL137492.1 |
- |
5 - 6 |
537 |
79 |
View Full Product Details