The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304782.1 | NP_001291711.1 | 2 - 3 | 323 | 75 |
NM_019604.3 | NP_062550.2 | 7 - 8 | 871 | 75 | |
XM_011542900.2 | XP_011541202.1 | 6 - 7 | 797 | 75 | |
GenBank mRNA | AB209830.1 | - | 2 - 3 | 232 | 75 |
AF001622.1 | - | 7 - 8 | 822 | 75 | |
AK293470.1 | - | 2 - 3 | 252 | 75 | |
AK313578.1 | - | 7 - 8 | 869 | 75 | |
BC070266.1 | - | 7 - 8 | 871 | 75 | |
DC305734.1 | - | 2 - 3 | 323 | 75 |