Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCMF1
Entrez Gene ID
56888
Gene Name
potassium channel modulatory factor 1
Gene Aliases
DEBT91, FIGC, PCMF, ZZZ1
Chromosome Location
Chr.2: 84971108 - 85059472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020122.4 NP_064507.3 5 - 6 947 67
XM_006712052.3 XP_006712115.1 5 - 6 979 67
XM_011532990.1 XP_011531292.1 5 - 6 869 67
XM_017004511.1 XP_016860000.1 4 - 5 583 67
GenBank mRNA AB083199.1 - 5 - 6 656 67
AF155652.1 - 5 - 6 656 67
AK023403.1 - 5 - 6 626 67
AK314761.1 - 5 - 6 722 67
AL122115.1 - 3 - 4 300 67
BC000178.2 - 5 - 6 746 67

View Full Product Details