Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38367227-38367773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERG
Entrez Gene ID
2078
Gene Name
ERG, ETS transcription factor
Gene Aliases
erg-3, p55
UniGene
Hs.473819
Gene Chromosome Location
Chr.21: 38367261-38662045 on Build GRCh38
Transcripts
NM_001243432.2

SNP Detail

SNP ID Chromosome Location
rs931174028 Chr.21: 38367435-38367435 on Build GRCh38
rs2836346 Chr.21: 38367743-38367743 on Build GRCh38
rs911274918 Chr.21: 38367352-38367352 on Build GRCh38
rs896016141 Chr.21: 38367702-38367702 on Build GRCh38
rs967303709 Chr.21: 38367259-38367259 on Build GRCh38
rs977077797 Chr.21: 38367349-38367349 on Build GRCh38
rs959701579 Chr.21: 38367620-38367620 on Build GRCh38
rs772963560 Chr.21: 38367360-38367360 on Build GRCh38
rs945785586 Chr.21: 38367277-38367277 on Build GRCh38
rs991167393 Chr.21: 38367631-38367631 on Build GRCh38
rs1019936219 Chr.21: 38367502-38367502 on Build GRCh38
rs752888248 Chr.21: 38367438-38367438 on Build GRCh38
rs959754300 Chr.21: 38367468-38367468 on Build GRCh38
rs1049160330 Chr.21: 38367437-38367437 on Build GRCh38
rs187145928 Chr.21: 38367678-38367678 on Build GRCh38
rs996812105 Chr.21: 38367527-38367527 on Build GRCh38
rs901258048 Chr.21: 38367707-38367707 on Build GRCh38
rs953306992 Chr.21: 38367547-38367547 on Build GRCh38
rs918592675 Chr.21: 38367595-38367595 on Build GRCh38
rs925825293 Chr.21: 38367544-38367544 on Build GRCh38
rs887213996 Chr.21: 38367448-38367448 on Build GRCh38
rs114419699 Chr.21: 38367563-38367563 on Build GRCh38
rs925802963 Chr.21: 38367303-38367303 on Build GRCh38
rs868694191 Chr.21: 38367534-38367534 on Build GRCh38
rs542661665 Chr.21: 38367422-38367422 on Build GRCh38
rs1034530667 Chr.21: 38367464-38367464 on Build GRCh38
rs1004319135 Chr.21: 38367456-38367456 on Build GRCh38
rs759411459 Chr.21: 38367602-38367602 on Build GRCh38
rs1050223607 Chr.21: 38367324-38367324 on Build GRCh38
rs1011519152 Chr.21: 38367554-38367554 on Build GRCh38
rs1029034078 Chr.21: 38367538-38367538 on Build GRCh38
rs997291378 Chr.21: 38367710-38367710 on Build GRCh38
rs914338445 Chr.21: 38367273-38367273 on Build GRCh38
rs62217874 Chr.21: 38367556-38367556 on Build GRCh38
rs915843201 Chr.21: 38367632-38367632 on Build GRCh38
rs369215913 Chr.21: 38367353-38367353 on Build GRCh38
rs979117316 Chr.21: 38367295-38367295 on Build GRCh38
rs146217369 Chr.21: 38367706-38367706 on Build GRCh38
rs939803114 Chr.21: 38367686-38367690 on Build GRCh38
rs888706520 Chr.21: 38367447-38367447 on Build GRCh38
rs557498207 Chr.21: 38367654-38367654 on Build GRCh38
rs760801356 Chr.21: 38367682-38367682 on Build GRCh38
rs141499194 Chr.21: 38367315-38367315 on Build GRCh38
rs575344149 Chr.21: 38367397-38367397 on Build GRCh38
rs1048397262 Chr.21: 38367665-38367665 on Build GRCh38
rs767504340 Chr.21: 38367555-38367555 on Build GRCh38
rs1041143598 Chr.21: 38367573-38367573 on Build GRCh38
rs759589915 Chr.21: 38367516-38367516 on Build GRCh38
rs533795413 Chr.21: 38367673-38367673 on Build GRCh38
rs967484467 Chr.21: 38367525-38367525 on Build GRCh38
rs770811156 Chr.21: 38367463-38367463 on Build GRCh38
rs376417021 Chr.21: 38367562-38367562 on Build GRCh38
rs912907842 Chr.21: 38367255-38367255 on Build GRCh38
rs571208714 Chr.21: 38367672-38367672 on Build GRCh38
rs573061215 Chr.21: 38367726-38367726 on Build GRCh38
rs994420984 Chr.21: 38367427-38367427 on Build GRCh38
rs774298615 Chr.21: 38367474-38367474 on Build GRCh38
rs978700604 Chr.21: 38367531-38367531 on Build GRCh38
rs762789111 Chr.21: 38367361-38367361 on Build GRCh38
rs894351858 Chr.21: 38367423-38367423 on Build GRCh38
rs767382517 Chr.21: 38367606-38367606 on Build GRCh38
rs1049735121 Chr.21: 38367714-38367714 on Build GRCh38
rs747745124 Chr.21: 38367320-38367320 on Build GRCh38
rs537681232 Chr.21: 38367601-38367601 on Build GRCh38

View Full Product Details