Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38381676-38382178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERG
Entrez Gene ID
2078
Gene Name
ERG, ETS transcription factor
Gene Aliases
erg-3, p55
UniGene
Hs.473819
Gene Chromosome Location
Chr.21: 38367261-38662045 on Build GRCh38
Transcripts
NM_001136154.1 NM_001136155.1 NM_001243428.1 NM_001243429.1 NM_001243432.2 NM_004449.4 NM_182918.3 XM_011529486.2 XM_017028287.1 XM_017028288.1 XM_017028289.1

SNP Detail

SNP ID Chromosome Location
rs934080767 Chr.21: 38381716-38381716 on Build GRCh38
rs557911294 Chr.21: 38381857-38381857 on Build GRCh38
rs887367975 Chr.21: 38381872-38381872 on Build GRCh38
rs924536753 Chr.21: 38381754-38381754 on Build GRCh38
rs770180818 Chr.21: 38381982-38381982 on Build GRCh38
rs773512930 Chr.21: 38382012-38382012 on Build GRCh38
rs977498753 Chr.21: 38381958-38381958 on Build GRCh38
rs894681948 Chr.21: 38381880-38381880 on Build GRCh38
rs531748305 Chr.21: 38381797-38381797 on Build GRCh38
rs145269203 Chr.21: 38381935-38381936 on Build GRCh38
rs1024031891 Chr.21: 38381944-38381944 on Build GRCh38
rs763482025 Chr.21: 38382147-38382147 on Build GRCh38
rs990099782 Chr.21: 38381989-38381989 on Build GRCh38
rs971500536 Chr.21: 38381703-38381703 on Build GRCh38
rs547558320 Chr.21: 38382100-38382100 on Build GRCh38
rs1047510354 Chr.21: 38382148-38382148 on Build GRCh38
rs1022571584 Chr.21: 38381770-38381770 on Build GRCh38
rs901237427 Chr.21: 38381841-38381841 on Build GRCh38
rs975887639 Chr.21: 38382069-38382069 on Build GRCh38
rs957258014 Chr.21: 38381984-38381984 on Build GRCh38
rs909524238 Chr.21: 38381941-38381941 on Build GRCh38
rs904116495 Chr.21: 38381787-38381787 on Build GRCh38
rs1013026284 Chr.21: 38381901-38381901 on Build GRCh38
rs910457624 Chr.21: 38382035-38382035 on Build GRCh38
rs1007292478 Chr.21: 38381768-38381768 on Build GRCh38
rs999742120 Chr.21: 38381789-38381789 on Build GRCh38
rs551755084 Chr.21: 38381798-38381798 on Build GRCh38
rs933820174 Chr.21: 38382004-38382004 on Build GRCh38
rs35187357 Chr.21: 38381945-38381945 on Build GRCh38
rs969772762 Chr.21: 38381854-38381854 on Build GRCh38
rs565895562 Chr.21: 38381747-38381747 on Build GRCh38
rs1017139742 Chr.21: 38381879-38381879 on Build GRCh38
rs527752427 Chr.21: 38381975-38381975 on Build GRCh38
rs563168788 Chr.21: 38381790-38381790 on Build GRCh38
rs776995645 Chr.21: 38381775-38381775 on Build GRCh38
rs934001777 Chr.21: 38381865-38381865 on Build GRCh38
rs796125187 Chr.21: 38382047-38382047 on Build GRCh38
rs73437665 Chr.21: 38381957-38381957 on Build GRCh38
rs922689215 Chr.21: 38382128-38382128 on Build GRCh38
rs569791112 Chr.21: 38381934-38381935 on Build GRCh38
rs534022480 Chr.21: 38382029-38382029 on Build GRCh38
rs1005765040 Chr.21: 38381874-38381874 on Build GRCh38
rs1039837332 Chr.21: 38381799-38381799 on Build GRCh38
rs977096673 Chr.21: 38381729-38381729 on Build GRCh38
rs183723337 Chr.21: 38381871-38381871 on Build GRCh38
rs1024460491 Chr.21: 38381852-38381852 on Build GRCh38
rs768958493 Chr.21: 38381760-38381760 on Build GRCh38

View Full Product Details