Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA10
Entrez Gene ID
56934
Gene Name
carbonic anhydrase 10
Gene Aliases
CA-RPX, CARPX, HUCEP-15
Chromosome Location
Chr.17: 51630313 - 52160017 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082533.1 NP_001076002.1 4 - 5 1394 82
NM_001082534.1 NP_001076003.1 4 - 5 959 82
NM_020178.4 NP_064563.1 3 - 4 1268 82
GenBank mRNA AB001597.1 - 3 - 4 282 82
AB036836.1 - 4 - 5 1184 82
AF288385.1 - 4 - 5 950 82
AK294652.1 - 2 - 3 639 82
AK313001.1 - 3 - 4 844 82
AL161812.1 - 3 - 4 284 82
AY358509.1 - 3 - 4 562 82
BC020577.1 - 4 - 5 926 82
BC029865.1 - 4 - 5 928 82
BC047456.1 - 3 - 4 1181 82
BC068462.1 - 3 - 4 1227 82
CR749607.1 - 4 - 5 730 82

View Full Product Details