The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007466.2 | NP_001007467.1 | 4 - 5 | 2082 | 66 |
NM_020245.4 | NP_064630.2 | 4 - 5 | 2082 | 66 | |
XM_011535946.1 | XP_011534248.1 | 5 - 6 | 2759 | 66 | |
XM_017011063.1 | XP_016866552.1 | 6 - 7 | 2908 | 66 | |
XM_017011064.1 | XP_016866553.1 | 7 - 8 | 3073 | 66 | |
XM_017011065.1 | XP_016866554.1 | 6 - 7 | 2826 | 66 | |
XM_017011066.1 | XP_016866555.1 | 5 - 6 | 2716 | 66 | |
XM_017011067.1 | XP_016866556.1 | 5 - 6 | 2809 | 66 | |
XM_017011068.1 | XP_016866557.1 | 5 - 6 | 2775 | 66 | |
XM_017011069.1 | XP_016866558.1 | 6 - 7 | 2911 | 66 | |
XM_017011070.1 | XP_016866559.1 | 5 - 6 | 2759 | 66 | |
XM_017011071.1 | XP_016866560.1 | 6 - 7 | 2911 | 66 | |
GenBank mRNA | AB037818.1 | - | 4 - 5 | 2082 | 66 |
AF219946.2 | - | 4 - 5 | 2082 | 66 | |
AF288480.1 | - | 4 - 5 | 2082 | 66 | |
AK304056.1 | - | 4 - 5 | 1297 | 66 | |
BC152476.1 | - | 4 - 5 | 2082 | 66 |