The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040194.1 | NP_001035284.1 | 2 - 3 | 185 | 90 |
NM_020350.4 | NP_065083.3 | 2 - 3 | 185 | 90 | |
GenBank mRNA | AF165187.1 | - | 2 - 3 | 61 | 90 |
AF312374.1 | - | 2 - 3 | 116 | 90 | |
BC011946.1 | - | 2 - 3 | 91 | 90 | |
BC017328.2 | - | 2 - 3 | 123 | 90 | |
BI117521.1 | - | 2 - 3 | 157 | 90 |