Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAN1C1
Entrez Gene ID
57134
Gene Name
mannosidase alpha class 1C member 1
Gene Aliases
HMIC, MAN1A3, MAN1C, pp6318
Chromosome Location
Chr.1: 25616820 - 25786207 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289010.1 NP_001275939.1 3 - 4 1085 69
NM_020379.3 NP_065112.1 3 - 4 1085 69
GenBank mRNA AB209275.1 - 3 - 4 433 69
AF261655.1 - 3 - 4 1085 69
AK128560.1 - 2 - 3 153 69
BC137017.1 - 3 - 4 891 69

View Full Product Details