Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REXO4
Entrez Gene ID
57109
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
REX4, XPMC2, XPMC2H
Chromosome Location
Chr.9: 133406058 - 133418172 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001279349.1 NP_001266278.1 5 - 6 884 125
NM_001279350.1 NP_001266279.1 7 - 8 1222 125
NM_001279351.1 NP_001266280.1 7 - 8 1083 125
NM_020385.3 NP_065118.2 7 - 8 1400 125
NR_103995.1 5 - 6 859 125
NR_103996.1 6 - 7 1053 125
GenBank mRNA AF273304.1 - 7 - 8 1348 125
AF295774.1 - 7 - 8 1348 125
AK000247.1 - 4 - 5 261 125
AK025493.1 - 7 - 8 1254 125
AK295982.1 - 7 - 8 1083 125
AK304547.1 - 7 - 8 1138 125
AK310164.1 - 5 - 6 762 125
AK314333.1 - 7 - 8 1314 125
AK315904.1 - 8 - 9 1073 125
AL136894.1 - 7 - 8 1331 125
BC009274.2 - 7 - 8 1303 125
BC071691.1 - 6 - 7 912 125

View Full Product Details