Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH2
Entrez Gene ID
57158
Gene Name
junctophilin 2
Gene Aliases
CMH17, JP-2, JP2
Chromosome Location
Chr.20: 44111697 - 44187578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175913.3 NP_787109.2 1 - 2 1251 62
GenBank mRNA AL132999.1 - 1 - 2 766 62

View Full Product Details