The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020649.2 | NP_065700.1 | 4 - 5 | 369 | 64 |
GenBank mRNA | AF174482.1 | - | 4 - 5 | 344 | 64 |
AF266479.2 | - | 4 - 5 | 300 | 64 | |
AK074560.1 | - | 4 - 5 | 369 | 64 | |
BC008937.2 | - | 4 - 5 | 266 | 64 | |
BC009376.2 | - | 4 - 5 | 322 | 64 | |
BC019289.1 | - | 4 - 5 | 266 | 64 |