The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300780.1 | NP_001287709.1 | 11 - 12 | 1875 | 125 |
NM_020796.4 | NP_065847.1 | 11 - 12 | 1875 | 125 | |
XM_005272042.3 | XP_005272099.2 | 11 - 12 | 1439 | 125 | |
XM_006714663.1 | XP_006714726.1 | 11 - 12 | 1774 | 125 | |
XM_017009675.1 | XP_016865164.1 | 11 - 12 | 1179 | 125 | |
XM_017009676.1 | XP_016865165.1 | 6 - 7 | 3726 | 125 | |
GenBank mRNA | AB037789.1 | - | 11 - 12 | 1344 | 125 |
AF279656.1 | - | 11 - 12 | 1752 | 125 | |
AK027501.1 | - | 2 - 3 | 186 | 125 | |
AK027867.1 | - | 9 - 10 | 1000 | 125 | |
AK315143.1 | - | 11 - 12 | 1294 | 125 | |
BC032619.1 | - | 11 - 12 | 1886 | 125 |