The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282987.1 | NP_001269916.1 | 5 - 6 | 573 | 90 |
NM_001282988.1 | NP_001269917.1 | 6 - 7 | 677 | 90 | |
NM_001282989.1 | NP_001269918.1 | 4 - 5 | 521 | 90 | |
NM_001282990.1 | NP_001269919.1 | 5 - 6 | 542 | 90 | |
NM_001282991.1 | NP_001269920.1 | 3 - 4 | 419 | 90 | |
NM_021004.3 | NP_066284.2 | 7 - 8 | 779 | 90 | |
XM_006720005.3 | XP_006720068.1 | 6 - 7 | 644 | 90 | |
GenBank mRNA | AB045131.1 | - | 7 - 8 | 804 | 90 |
AF044127.1 | - | 7 - 8 | 750 | 90 | |
AF064256.1 | - | 7 - 8 | 753 | 90 | |
AK001870.1 | - | 7 - 8 | 745 | 90 | |
AK314448.1 | - | 7 - 8 | 744 | 90 | |
AY071856.1 | - | 4 - 5 | 472 | 90 | |
AY358638.1 | - | 7 - 8 | 760 | 90 | |
AY616182.1 | - | 5 - 6 | 573 | 90 | |
BC003019.1 | - | 7 - 8 | 727 | 90 | |
CA438339.1 | - | 1 - 2 | 90 | ||
CD674505.1 | - | 3 - 4 | 391 | 90 | |
DQ325464.1 | - | 6 - 7 | 644 | 90 | |
DQ338571.1 | - | 4 - 5 | 371 | 90 | |
DQ344810.1 | - | 7 - 8 | 758 | 90 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277864.1 | NP_001264793.1 | 4 - 5 | 912 | 90 |
NR_102687.1 | 3 - 4 | 647 | 90 | ||
NR_102688.1 | 4 - 5 | 825 | 90 | ||
NR_102689.1 | 4 - 5 | 589 | 90 | ||
NR_102691.1 | 3 - 4 | 411 | 90 | ||
NR_102692.1 | 4 - 5 | 672 | 90 | ||
NR_102693.1 | 5 - 6 | 768 | 90 | ||
NR_102694.1 | 3 - 4 | 537 | 90 | ||
XM_006720246.3 | XP_006720309.1 | 6 - 7 | 640 | 90 | |
GenBank mRNA | BC171914.1 | - | 4 - 5 | 631 | 90 |
BC171918.1 | - | 5 - 6 | 727 | 90 | |
GQ871922.1 | - | 3 - 4 | 384 | 90 | |
GQ871923.1 | - | 4 - 5 | 588 | 90 | |
GQ871924.1 | - | 4 - 5 | 861 | 90 | |
KU058702.1 | - | 4 - 5 | 849 | 90 | |
KU058703.1 | - | 4 - 5 | 788 | 90 | |
KU058704.1 | - | 4 - 5 | 766 | 90 | |
KU058705.1 | - | 4 - 5 | 716 | 90 | |
KU058706.1 | - | 3 - 4 | 714 | 90 | |
KU058707.1 | - | 3 - 4 | 588 | 90 | |
KU058708.1 | - | 4 - 5 | 570 | 90 | |
KU058709.1 | - | 2 - 3 | 453 | 90 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193635.1 | NP_001180564.1 | 8 - 9 | 812 | 90 |
NM_001193636.1 | NP_001180565.1 | 7 - 8 | 1055 | 90 | |
NM_001193637.1 | NP_001180566.1 | 5 - 6 | 932 | 90 | |
NM_198083.3 | NP_932349.2 | 7 - 8 | 854 | 90 | |
GenBank mRNA | AI087304.1 | - | 1 - 2 | 90 | |
AY616183.1 | - | 3 - 4 | 733 | 90 | |
AY943857.1 | - | 3 - 4 | 631 | 90 | |
BC000663.2 | - | 7 - 8 | 762 | 90 | |
DN237881.1 | - | 5 - 6 | 618 | 90 | |
DN237882.1 | - | 5 - 6 | 539 | 90 | |
DQ088987.1 | - | 5 - 6 | 353 | 90 | |
GU564234.1 | - | 8 - 9 | 721 | 90 | |
KC237375.1 | - | 5 - 6 | 851 | 90 | |
KU141377.1 | - | 5 - 6 | 825 | 90 |