The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021079.4 | NP_066565.1 | 3 - 4 | 419 | 111 |
XM_011524858.2 | XP_011523160.1 | 3 - 4 | 427 | 111 | |
GenBank mRNA | AF020500.1 | - | 3 - 4 | 375 | 111 |
AF043324.1 | - | 3 - 4 | 398 | 111 | |
AK291936.1 | - | 3 - 4 | 398 | 111 | |
BC006538.2 | - | 3 - 4 | 406 | 111 | |
BC006569.2 | - | 3 - 4 | 406 | 111 | |
BC007258.2 | - | 2 - 3 | 253 | 111 | |
BC008312.2 | - | 2 - 3 | 253 | 111 | |
BC008579.1 | - | 3 - 4 | 382 | 111 | |
BU849351.1 | - | 3 - 4 | 419 | 111 | |
M86707.1 | - | 2 - 3 | 238 | 111 |