Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25564529-25565056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK3
Entrez Gene ID
157
Gene Name
G protein-coupled receptor kinase 3
Gene Aliases
ADRBK2, BARK2
UniGene
Hs.657494
Gene Chromosome Location
Chr.22: 25564894-25729294 on Build GRCh38
Transcripts
NM_005160.3

SNP Detail

SNP ID Chromosome Location
rs1011694013 Chr.22: 25564925-25564925 on Build GRCh38
rs1044231075 Chr.22: 25564886-25564886 on Build GRCh38
rs948165991 Chr.22: 25564968-25564969 on Build GRCh38
rs111767462 Chr.22: 25564908-25564908 on Build GRCh38
rs1044177155 Chr.22: 25564975-25564975 on Build GRCh38
rs1039312926 Chr.22: 25564573-25564573 on Build GRCh38
rs907888117 Chr.22: 25564791-25564791 on Build GRCh38
rs1022827338 Chr.22: 25564674-25564674 on Build GRCh38
rs1053036371 Chr.22: 25564984-25564984 on Build GRCh38
rs913665539 Chr.22: 25564895-25564913 on Build GRCh38
rs907050691 Chr.22: 25564977-25564977 on Build GRCh38
rs1040971985 Chr.22: 25564910-25564910 on Build GRCh38
rs942032273 Chr.22: 25564950-25564950 on Build GRCh38
rs1031717944 Chr.22: 25564901-25564902 on Build GRCh38
rs989920741 Chr.22: 25564738-25564738 on Build GRCh38
rs780157517 Chr.22: 25564999-25564999 on Build GRCh38
rs964481505 Chr.22: 25564930-25564930 on Build GRCh38
rs923499277 Chr.22: 25564768-25564768 on Build GRCh38
rs945093343 Chr.22: 25564898-25564908 on Build GRCh38
rs529474941 Chr.22: 25564924-25564924 on Build GRCh38
rs964868213 Chr.22: 25564739-25564739 on Build GRCh38
rs971324802 Chr.22: 25564680-25564680 on Build GRCh38
rs990030088 Chr.22: 25564886-25564911 on Build GRCh38
rs1038586838 Chr.22: 25564636-25564636 on Build GRCh38
rs116581378 Chr.22: 25564552-25564552 on Build GRCh38
rs983723153 Chr.22: 25564947-25564947 on Build GRCh38
rs1021383516 Chr.22: 25564929-25564929 on Build GRCh38
rs898335792 Chr.22: 25564644-25564644 on Build GRCh38
rs1031362194 Chr.22: 25564717-25564717 on Build GRCh38
rs942070178 Chr.22: 25564819-25564819 on Build GRCh38
rs914767642 Chr.22: 25564554-25564555 on Build GRCh38
rs541169558 Chr.22: 25565019-25565019 on Build GRCh38
rs1006319408 Chr.22: 25564634-25564634 on Build GRCh38
rs755536380 Chr.22: 25565034-25565034 on Build GRCh38
rs981795140 Chr.22: 25564962-25564963 on Build GRCh38
rs955961740 Chr.22: 25564737-25564737 on Build GRCh38
rs895267246 Chr.22: 25564851-25564851 on Build GRCh38
rs923446905 Chr.22: 25564939-25564939 on Build GRCh38
rs41261045 Chr.22: 25564658-25564658 on Build GRCh38
rs753901708 Chr.22: 25565022-25565022 on Build GRCh38
rs559915130 Chr.22: 25565035-25565035 on Build GRCh38
rs974948029 Chr.22: 25564751-25564751 on Build GRCh38
rs1047747574 Chr.22: 25564610-25564610 on Build GRCh38
rs933590275 Chr.22: 25564777-25564777 on Build GRCh38
rs867796856 Chr.22: 25564946-25564946 on Build GRCh38
rs576349029 Chr.22: 25564563-25564563 on Build GRCh38
rs1037798239 Chr.22: 25564839-25564839 on Build GRCh38
rs775331281 Chr.22: 25565011-25565013 on Build GRCh38
rs973849854 Chr.22: 25564952-25564952 on Build GRCh38
rs900224452 Chr.22: 25565001-25565001 on Build GRCh38
rs938566352 Chr.22: 25564983-25564983 on Build GRCh38
rs756094047 Chr.22: 25564993-25564993 on Build GRCh38
rs886510347 Chr.22: 25564618-25564618 on Build GRCh38
rs576935980 Chr.22: 25564706-25564706 on Build GRCh38
rs958233208 Chr.22: 25564585-25564614 on Build GRCh38
rs1048164326 Chr.22: 25564780-25564780 on Build GRCh38
rs571997613 Chr.22: 25564572-25564574 on Build GRCh38
rs749077845 Chr.22: 25565010-25565011 on Build GRCh38
rs923553079 Chr.22: 25564581-25564583 on Build GRCh38
rs180999646 Chr.22: 25564693-25564693 on Build GRCh38
rs955105384 Chr.22: 25564945-25564946 on Build GRCh38

View Full Product Details