Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25672187-25672694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK3
Entrez Gene ID
157
Gene Name
G protein-coupled receptor kinase 3
Gene Aliases
ADRBK2, BARK2
UniGene
Hs.657494
Gene Chromosome Location
Chr.22: 25564894-25729294 on Build GRCh38
Transcripts
NM_005160.3 XM_011529975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244561 c.507delC p.D169fs*14 Chr.22: 25672299-25672299 on Build GRCh38
1032806 c.537C>T p.N179N Chr.22: 25672329-25672329 on Build GRCh38
5821378 c.517A>T p.R173* Chr.22: 25672309-25672309 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979300672 Chr.22: 25672618-25672618 on Build GRCh38
rs780557371 Chr.22: 25672313-25672313 on Build GRCh38
rs200047025 Chr.22: 25672279-25672279 on Build GRCh38
rs115146499 Chr.22: 25672534-25672534 on Build GRCh38
rs889279149 Chr.22: 25672301-25672301 on Build GRCh38
rs916121230 Chr.22: 25672572-25672572 on Build GRCh38
rs867447127 Chr.22: 25672299-25672299 on Build GRCh38
rs761190491 Chr.22: 25672364-25672364 on Build GRCh38
rs772648348 Chr.22: 25672375-25672375 on Build GRCh38
rs968923247 Chr.22: 25672595-25672595 on Build GRCh38
rs771636167 Chr.22: 25672368-25672368 on Build GRCh38
rs757388758 Chr.22: 25672262-25672262 on Build GRCh38
rs202119424 Chr.22: 25672287-25672287 on Build GRCh38
rs145784362 Chr.22: 25672629-25672629 on Build GRCh38
rs1019085021 Chr.22: 25672345-25672345 on Build GRCh38
rs141366731 Chr.22: 25672475-25672475 on Build GRCh38
rs553542402 Chr.22: 25672215-25672215 on Build GRCh38
rs565783706 Chr.22: 25672229-25672229 on Build GRCh38
rs1028037833 Chr.22: 25672441-25672441 on Build GRCh38
rs543028680 Chr.22: 25672448-25672448 on Build GRCh38
rs757029864 Chr.22: 25672256-25672256 on Build GRCh38
rs570592605 Chr.22: 25672211-25672211 on Build GRCh38
rs559936561 Chr.22: 25672565-25672565 on Build GRCh38
rs145041542 Chr.22: 25672493-25672493 on Build GRCh38
rs917105563 Chr.22: 25672600-25672601 on Build GRCh38
rs1050315501 Chr.22: 25672658-25672658 on Build GRCh38
rs751684824 Chr.22: 25672267-25672267 on Build GRCh38
rs781500942 Chr.22: 25672270-25672270 on Build GRCh38
rs5761150 Chr.22: 25672525-25672525 on Build GRCh38
rs952113213 Chr.22: 25672446-25672446 on Build GRCh38
rs924802875 Chr.22: 25672625-25672625 on Build GRCh38
rs1016022588 Chr.22: 25672439-25672439 on Build GRCh38
rs774153928 Chr.22: 25672354-25672354 on Build GRCh38
rs542477953 Chr.22: 25672636-25672636 on Build GRCh38
rs958516408 Chr.22: 25672509-25672509 on Build GRCh38
rs888972969 Chr.22: 25672432-25672432 on Build GRCh38
rs1041095842 Chr.22: 25672630-25672630 on Build GRCh38
rs1035387659 Chr.22: 25672484-25672484 on Build GRCh38
rs1041752740 Chr.22: 25672248-25672248 on Build GRCh38
rs369418687 Chr.22: 25672282-25672282 on Build GRCh38
rs904191487 Chr.22: 25672255-25672255 on Build GRCh38
rs1006437685 Chr.22: 25672436-25672436 on Build GRCh38
rs112818401 Chr.22: 25672613-25672613 on Build GRCh38
rs924657861 Chr.22: 25672261-25672261 on Build GRCh38
rs932630866 Chr.22: 25672642-25672642 on Build GRCh38
rs182942871 Chr.22: 25672212-25672212 on Build GRCh38
rs73167000 Chr.22: 25672330-25672330 on Build GRCh38
rs749724802 Chr.22: 25672329-25672329 on Build GRCh38

View Full Product Details