Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25678674-25679175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK3
Entrez Gene ID
157
Gene Name
G protein-coupled receptor kinase 3
Gene Aliases
ADRBK2, BARK2
UniGene
Hs.657494
Gene Chromosome Location
Chr.22: 25564894-25729294 on Build GRCh38
Transcripts
NM_005160.3 XM_011529975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032810 c.659A>C p.K220T Chr.22: 25678827-25678827 on Build GRCh38
4660153 c.745G>A p.G249R Chr.22: 25678913-25678913 on Build GRCh38
6053185 c.731C>G p.S244C Chr.22: 25678899-25678899 on Build GRCh38
4556600 c.697G>A p.E233K Chr.22: 25678865-25678865 on Build GRCh38
6094940 c.747+1G>C p.? Chr.22: 25678916-25678916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976291225 Chr.22: 25679022-25679022 on Build GRCh38
rs374160084 Chr.22: 25678783-25678783 on Build GRCh38
rs769787425 Chr.22: 25678795-25678795 on Build GRCh38
rs199846039 Chr.22: 25678758-25678758 on Build GRCh38
rs754013509 Chr.22: 25678918-25678918 on Build GRCh38
rs765722291 Chr.22: 25678945-25678945 on Build GRCh38
rs1044802 Chr.22: 25678876-25678876 on Build GRCh38
rs3730108 Chr.22: 25678773-25678773 on Build GRCh38
rs779921948 Chr.22: 25678906-25678910 on Build GRCh38
rs749092777 Chr.22: 25678957-25678957 on Build GRCh38
rs376664010 Chr.22: 25678965-25678965 on Build GRCh38
rs745771303 Chr.22: 25678793-25678793 on Build GRCh38
rs148213407 Chr.22: 25679078-25679078 on Build GRCh38
rs760791450 Chr.22: 25678881-25678881 on Build GRCh38
rs770447748 Chr.22: 25678786-25678786 on Build GRCh38
rs769607201 Chr.22: 25679114-25679114 on Build GRCh38
rs2877289 Chr.22: 25678744-25678744 on Build GRCh38
rs941814200 Chr.22: 25679113-25679113 on Build GRCh38
rs758057650 Chr.22: 25678775-25678775 on Build GRCh38
rs766547650 Chr.22: 25678893-25678893 on Build GRCh38
rs919486925 Chr.22: 25679026-25679026 on Build GRCh38
rs3730109 Chr.22: 25679039-25679039 on Build GRCh38
rs1031684823 Chr.22: 25678975-25678975 on Build GRCh38
rs1049059505 Chr.22: 25679038-25679038 on Build GRCh38
rs1055639721 Chr.22: 25679090-25679090 on Build GRCh38
rs903068375 Chr.22: 25679130-25679130 on Build GRCh38
rs1023113606 Chr.22: 25678859-25678859 on Build GRCh38
rs986318413 Chr.22: 25678865-25678865 on Build GRCh38
rs756014658 Chr.22: 25678893-25678895 on Build GRCh38
rs375385425 Chr.22: 25678722-25678722 on Build GRCh38
rs142131104 Chr.22: 25678721-25678721 on Build GRCh38
rs968837036 Chr.22: 25678932-25678932 on Build GRCh38
rs575157082 Chr.22: 25678952-25678952 on Build GRCh38
rs199789488 Chr.22: 25678812-25678812 on Build GRCh38
rs185263910 Chr.22: 25679063-25679063 on Build GRCh38
rs1050305646 Chr.22: 25678701-25678701 on Build GRCh38
rs755538776 Chr.22: 25678920-25678920 on Build GRCh38
rs201402565 Chr.22: 25678782-25678782 on Build GRCh38
rs910405014 Chr.22: 25679092-25679092 on Build GRCh38
rs776166527 Chr.22: 25678791-25678791 on Build GRCh38
rs957083772 Chr.22: 25678836-25678836 on Build GRCh38
rs559771760 Chr.22: 25679128-25679128 on Build GRCh38
rs758719712 Chr.22: 25678955-25678955 on Build GRCh38
rs929041404 Chr.22: 25679139-25679139 on Build GRCh38
rs565437355 Chr.22: 25679046-25679046 on Build GRCh38
rs956238900 Chr.22: 25678994-25678994 on Build GRCh38
rs773341448 Chr.22: 25678829-25678829 on Build GRCh38
rs976816494 Chr.22: 25679034-25679034 on Build GRCh38
rs954676120 Chr.22: 25679041-25679041 on Build GRCh38
rs998735632 Chr.22: 25679144-25679144 on Build GRCh38
rs200189647 Chr.22: 25678954-25678954 on Build GRCh38
rs201479633 Chr.22: 25678806-25678806 on Build GRCh38

View Full Product Details