Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26450961-26451471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_011530492.1 XM_017029045.1 XM_017029048.1 XM_017029049.1

SNP Detail

SNP ID Chromosome Location
rs558267465 Chr.22: 26451326-26451326 on Build GRCh38
rs918060804 Chr.22: 26451222-26451222 on Build GRCh38
rs186245688 Chr.22: 26451313-26451313 on Build GRCh38
rs143168600 Chr.22: 26451383-26451383 on Build GRCh38
rs913043974 Chr.22: 26451201-26451201 on Build GRCh38
rs889464815 Chr.22: 26451348-26451348 on Build GRCh38
rs746034881 Chr.22: 26451221-26451221 on Build GRCh38
rs139365115 Chr.22: 26451210-26451210 on Build GRCh38
rs531060820 Chr.22: 26451167-26451167 on Build GRCh38
rs777259855 Chr.22: 26451023-26451024 on Build GRCh38
rs988617862 Chr.22: 26451197-26451197 on Build GRCh38
rs536794522 Chr.22: 26451321-26451321 on Build GRCh38
rs751954729 Chr.22: 26451054-26451054 on Build GRCh38
rs374464989 Chr.22: 26451231-26451231 on Build GRCh38
rs995026603 Chr.22: 26451031-26451031 on Build GRCh38
rs565702698 Chr.22: 26451217-26451217 on Build GRCh38
rs957303657 Chr.22: 26451439-26451439 on Build GRCh38
rs536065020 Chr.22: 26451226-26451226 on Build GRCh38
rs773217565 Chr.22: 26451384-26451384 on Build GRCh38
rs1000969308 Chr.22: 26450990-26450990 on Build GRCh38
rs761998254 Chr.22: 26451103-26451104 on Build GRCh38
rs554045554 Chr.22: 26451213-26451213 on Build GRCh38
rs764578879 Chr.22: 26451042-26451042 on Build GRCh38
rs548671005 Chr.22: 26451045-26451045 on Build GRCh38
rs5761533 Chr.22: 26451271-26451271 on Build GRCh38
rs957504326 Chr.22: 26451184-26451184 on Build GRCh38
rs147511078 Chr.22: 26451443-26451443 on Build GRCh38
rs972646133 Chr.22: 26451088-26451088 on Build GRCh38
rs969716838 Chr.22: 26451096-26451096 on Build GRCh38
rs1032074143 Chr.22: 26450991-26450991 on Build GRCh38
rs1010068625 Chr.22: 26451447-26451447 on Build GRCh38
rs953817375 Chr.22: 26451012-26451012 on Build GRCh38
rs1006607051 Chr.22: 26451354-26451354 on Build GRCh38
rs1053899508 Chr.22: 26451332-26451332 on Build GRCh38
rs1035029671 Chr.22: 26451208-26451208 on Build GRCh38
rs114419177 Chr.22: 26451424-26451424 on Build GRCh38
rs543210717 Chr.22: 26451285-26451285 on Build GRCh38
rs756332446 Chr.22: 26451230-26451230 on Build GRCh38
rs1019394656 Chr.22: 26451371-26451371 on Build GRCh38
rs757502636 Chr.22: 26451155-26451155 on Build GRCh38
rs1013987506 Chr.22: 26451062-26451062 on Build GRCh38
rs1033062885 Chr.22: 26451431-26451431 on Build GRCh38
rs1032968679 Chr.22: 26451181-26451181 on Build GRCh38
rs565814780 Chr.22: 26451087-26451087 on Build GRCh38
rs377464803 Chr.22: 26451270-26451270 on Build GRCh38
rs552595828 Chr.22: 26451205-26451205 on Build GRCh38
rs150263442 Chr.22: 26451024-26451024 on Build GRCh38
rs575718170 Chr.22: 26451314-26451314 on Build GRCh38
rs557130630 Chr.22: 26451324-26451324 on Build GRCh38
rs35133132 Chr.22: 26451221-26451222 on Build GRCh38
rs920883442 Chr.22: 26451229-26451229 on Build GRCh38
rs770192250 Chr.22: 26451373-26451373 on Build GRCh38
rs994429401 Chr.22: 26451413-26451413 on Build GRCh38
rs1026469506 Chr.22: 26451032-26451032 on Build GRCh38
rs866461994 Chr.22: 26451228-26451228 on Build GRCh38
rs141992750 Chr.22: 26451017-26451017 on Build GRCh38
rs570870645 Chr.22: 26451207-26451207 on Build GRCh38

View Full Product Details