Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26452523-26453034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_006724353.3 XM_006724354.3 XM_006724360.3 XM_011530485.1 XM_011530486.2 XM_011530487.2 XM_011530488.2 XM_011530489.2 XM_011530490.2 XM_011530492.1 XM_011530493.2 XM_011530494.2 XM_011530495.2 XM_011530496.2 XM_017029042.1 XM_017029043.1 XM_017029044.1 XM_017029045.1 XM_017029046.1 XM_017029047.1 XM_017029048.1 XM_017029049.1 XM_017029050.1 XM_017029051.1 XM_017029052.1 XM_017029053.1 XM_017029054.1 XM_017029055.1 XM_017029056.1 XM_017029057.1 XM_017029058.1 XM_017029059.1 XM_017029060.1 XM_017029061.1 XM_017029062.1 XM_017029063.1 XM_017029064.1

SNP Detail

SNP ID Chromosome Location
rs1011743668 Chr.22: 26452765-26452765 on Build GRCh38
rs758700168 Chr.22: 26452786-26452786 on Build GRCh38
rs886057314 Chr.22: 26452718-26452718 on Build GRCh38
rs1034327772 Chr.22: 26452761-26452761 on Build GRCh38
rs899956428 Chr.22: 26452795-26452795 on Build GRCh38
rs746883110 Chr.22: 26452633-26452633 on Build GRCh38
rs912323680 Chr.22: 26452820-26452820 on Build GRCh38
rs1039791914 Chr.22: 26452782-26452782 on Build GRCh38
rs991140929 Chr.22: 26452922-26452922 on Build GRCh38
rs182700538 Chr.22: 26453005-26453005 on Build GRCh38
rs919538491 Chr.22: 26452630-26452630 on Build GRCh38
rs962373521 Chr.22: 26452646-26452646 on Build GRCh38
rs965143883 Chr.22: 26452821-26452821 on Build GRCh38
rs886908582 Chr.22: 26452708-26452708 on Build GRCh38
rs1046218756 Chr.22: 26453002-26453002 on Build GRCh38
rs565394140 Chr.22: 26452671-26452671 on Build GRCh38
rs909511730 Chr.22: 26452646-26452650 on Build GRCh38
rs1004445896 Chr.22: 26452859-26452859 on Build GRCh38
rs938758417 Chr.22: 26453009-26453009 on Build GRCh38
rs548085200 Chr.22: 26452615-26452615 on Build GRCh38
rs775062349 Chr.22: 26452770-26452770 on Build GRCh38
rs570509591 Chr.22: 26452753-26452753 on Build GRCh38
rs1017155909 Chr.22: 26452869-26452869 on Build GRCh38
rs568066326 Chr.22: 26452930-26452930 on Build GRCh38
rs977104076 Chr.22: 26452563-26452563 on Build GRCh38
rs908457988 Chr.22: 26452987-26452987 on Build GRCh38
rs773328400 Chr.22: 26452796-26452796 on Build GRCh38
rs552113955 Chr.22: 26452740-26452740 on Build GRCh38
rs967101864 Chr.22: 26452561-26452561 on Build GRCh38
rs951634381 Chr.22: 26452857-26452857 on Build GRCh38
rs1021754829 Chr.22: 26452769-26452769 on Build GRCh38
rs1048376204 Chr.22: 26452898-26452898 on Build GRCh38
rs959233919 Chr.22: 26452764-26452764 on Build GRCh38
rs763552637 Chr.22: 26452975-26452975 on Build GRCh38
rs745672037 Chr.22: 26452714-26452714 on Build GRCh38
rs552928725 Chr.22: 26452810-26452810 on Build GRCh38
rs951696387 Chr.22: 26452634-26452634 on Build GRCh38
rs971085046 Chr.22: 26452953-26452953 on Build GRCh38
rs1029797277 Chr.22: 26452855-26452855 on Build GRCh38
rs531146121 Chr.22: 26452974-26452974 on Build GRCh38
rs1048260688 Chr.22: 26452692-26452692 on Build GRCh38
rs939860499 Chr.22: 26453001-26453001 on Build GRCh38
rs867158897 Chr.22: 26452604-26452604 on Build GRCh38
rs886057316 Chr.22: 26452864-26452864 on Build GRCh38
rs762469062 Chr.22: 26452939-26452939 on Build GRCh38
rs536866354 Chr.22: 26452705-26452705 on Build GRCh38
rs920914838 Chr.22: 26452882-26452882 on Build GRCh38
rs1003317798 Chr.22: 26452757-26452757 on Build GRCh38
rs76496430 Chr.22: 26452756-26452756 on Build GRCh38
rs1036488000 Chr.22: 26452732-26452732 on Build GRCh38
rs983447267 Chr.22: 26452636-26452636 on Build GRCh38
rs886057315 Chr.22: 26452842-26452842 on Build GRCh38
rs1004041123 Chr.22: 26452719-26452719 on Build GRCh38
rs931226935 Chr.22: 26452883-26452883 on Build GRCh38
rs931188986 Chr.22: 26452686-26452686 on Build GRCh38
rs776345185 Chr.22: 26452713-26452713 on Build GRCh38
rs36046278 Chr.22: 26452810-26452811 on Build GRCh38
rs981599986 Chr.22: 26452982-26452982 on Build GRCh38
rs912304266 Chr.22: 26452593-26452593 on Build GRCh38
rs915693261 Chr.22: 26452683-26452683 on Build GRCh38
rs879690761 Chr.22: 26452868-26452868 on Build GRCh38
rs975497185 Chr.22: 26452864-26452864 on Build GRCh38

View Full Product Details