Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26453200-26453699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_006724353.3 XM_006724354.3 XM_006724360.3 XM_011530485.1 XM_011530486.2 XM_011530487.2 XM_011530488.2 XM_011530489.2 XM_011530490.2 XM_011530491.2 XM_011530492.1 XM_011530493.2 XM_011530494.2 XM_011530495.2 XM_011530496.2 XM_017029042.1 XM_017029043.1 XM_017029044.1 XM_017029045.1 XM_017029046.1 XM_017029047.1 XM_017029048.1 XM_017029049.1 XM_017029050.1 XM_017029051.1 XM_017029052.1 XM_017029053.1 XM_017029054.1 XM_017029055.1 XM_017029056.1 XM_017029057.1 XM_017029058.1 XM_017029059.1 XM_017029060.1 XM_017029061.1 XM_017029062.1 XM_017029063.1 XM_017029064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5519792 c.2020G>A p.A674T Chr.22: 26453340-26453340 on Build GRCh38
4103175 c.2097G>A p.Q699Q Chr.22: 26453302-26453302 on Build GRCh38
1032962 c.2081G>A p.G694D Chr.22: 26453279-26453279 on Build GRCh38
212943 c.2041G>A p.G681S Chr.22: 26453319-26453319 on Build GRCh38
6681971 c.2016C>T p.Y672Y Chr.22: 26453383-26453383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761967529 Chr.22: 26453392-26453392 on Build GRCh38
rs955879389 Chr.22: 26453389-26453389 on Build GRCh38
rs756494145 Chr.22: 26453335-26453335 on Build GRCh38
rs281865100 Chr.22: 26453266-26453267 on Build GRCh38
rs898667711 Chr.22: 26453478-26453478 on Build GRCh38
rs570600548 Chr.22: 26453645-26453645 on Build GRCh38
rs746961193 Chr.22: 26453356-26453356 on Build GRCh38
rs752931351 Chr.22: 26453320-26453320 on Build GRCh38
rs997421605 Chr.22: 26453510-26453510 on Build GRCh38
rs146737414 Chr.22: 26453660-26453660 on Build GRCh38
rs776239550 Chr.22: 26453372-26453372 on Build GRCh38
rs867819224 Chr.22: 26453390-26453390 on Build GRCh38
rs768058057 Chr.22: 26453310-26453310 on Build GRCh38
rs761348469 Chr.22: 26453304-26453304 on Build GRCh38
rs982014853 Chr.22: 26453579-26453579 on Build GRCh38
rs367897224 Chr.22: 26453250-26453250 on Build GRCh38
rs927781508 Chr.22: 26453374-26453374 on Build GRCh38
rs927860701 Chr.22: 26453604-26453604 on Build GRCh38
rs1028097963 Chr.22: 26453511-26453511 on Build GRCh38
rs1013096474 Chr.22: 26453537-26453537 on Build GRCh38
rs767769111 Chr.22: 26453393-26453393 on Build GRCh38
rs144248515 Chr.22: 26453383-26453383 on Build GRCh38
rs138189133 Chr.22: 26453281-26453281 on Build GRCh38
rs769525247 Chr.22: 26453624-26453624 on Build GRCh38
rs567467720 Chr.22: 26453242-26453242 on Build GRCh38
rs750947119 Chr.22: 26453332-26453332 on Build GRCh38
rs187997943 Chr.22: 26453429-26453429 on Build GRCh38
rs937897105 Chr.22: 26453618-26453618 on Build GRCh38
rs374200129 Chr.22: 26453348-26453348 on Build GRCh38
rs148294572 Chr.22: 26453394-26453394 on Build GRCh38
rs371133967 Chr.22: 26453445-26453445 on Build GRCh38
rs770063857 Chr.22: 26453271-26453271 on Build GRCh38
rs940203057 Chr.22: 26453438-26453438 on Build GRCh38
rs773131180 Chr.22: 26453350-26453350 on Build GRCh38
rs757412572 Chr.22: 26453327-26453327 on Build GRCh38
rs368602340 Chr.22: 26453360-26453360 on Build GRCh38
rs202120615 Chr.22: 26453401-26453401 on Build GRCh38
rs1038464720 Chr.22: 26453457-26453457 on Build GRCh38
rs768192969 Chr.22: 26453295-26453295 on Build GRCh38
rs781772371 Chr.22: 26453251-26453251 on Build GRCh38
rs952428609 Chr.22: 26453518-26453518 on Build GRCh38
rs781116258 Chr.22: 26453493-26453493 on Build GRCh38
rs768278011 Chr.22: 26453673-26453673 on Build GRCh38
rs983803544 Chr.22: 26453526-26453526 on Build GRCh38
rs754550717 Chr.22: 26453339-26453339 on Build GRCh38
rs376367198 Chr.22: 26453623-26453623 on Build GRCh38
rs906853794 Chr.22: 26453553-26453553 on Build GRCh38
rs747600132 Chr.22: 26453345-26453345 on Build GRCh38
rs145698999 Chr.22: 26453269-26453269 on Build GRCh38
rs770100093 Chr.22: 26453620-26453620 on Build GRCh38
rs748711601 Chr.22: 26453614-26453614 on Build GRCh38
rs189953957 Chr.22: 26453436-26453436 on Build GRCh38
rs777195255 Chr.22: 26453431-26453431 on Build GRCh38
rs9625029 Chr.22: 26453398-26453398 on Build GRCh38
rs370493873 Chr.22: 26453323-26453323 on Build GRCh38
rs757158956 Chr.22: 26453420-26453421 on Build GRCh38
rs764344335 Chr.22: 26453380-26453380 on Build GRCh38
rs747863960 Chr.22: 26453430-26453430 on Build GRCh38
rs760506995 Chr.22: 26453314-26453314 on Build GRCh38
rs771929646 Chr.22: 26453307-26453307 on Build GRCh38
rs761057923 Chr.22: 26453324-26453324 on Build GRCh38
rs752806729 Chr.22: 26453468-26453468 on Build GRCh38
rs752827715 Chr.22: 26453392-26453393 on Build GRCh38
rs755354422 Chr.22: 26453417-26453417 on Build GRCh38
rs757207651 Chr.22: 26453474-26453474 on Build GRCh38
rs779739799 Chr.22: 26453280-26453280 on Build GRCh38
rs759405900 Chr.22: 26453373-26453373 on Build GRCh38
rs921242818 Chr.22: 26453424-26453424 on Build GRCh38
rs754088753 Chr.22: 26453407-26453407 on Build GRCh38
rs773804490 Chr.22: 26453299-26453299 on Build GRCh38
rs530118832 Chr.22: 26453382-26453382 on Build GRCh38
rs1015247708 Chr.22: 26453531-26453531 on Build GRCh38
rs558103922 Chr.22: 26453231-26453231 on Build GRCh38
rs768075866 Chr.22: 26453328-26453328 on Build GRCh38
rs563217135 Chr.22: 26453559-26453559 on Build GRCh38
rs201206642 Chr.22: 26453400-26453400 on Build GRCh38
rs1044639211 Chr.22: 26453545-26453545 on Build GRCh38
rs749689033 Chr.22: 26453336-26453336 on Build GRCh38
rs373741660 Chr.22: 26453228-26453228 on Build GRCh38
rs779709959 Chr.22: 26453637-26453637 on Build GRCh38
rs778546132 Chr.22: 26453344-26453344 on Build GRCh38
rs552338204 Chr.22: 26453632-26453632 on Build GRCh38
rs149568178 Chr.22: 26453319-26453319 on Build GRCh38

View Full Product Details