Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26617736-26618249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBA4
Entrez Gene ID
1413
Gene Name
crystallin beta A4
Gene Aliases
CTRCT23, CYRBA4, MCOPCT4
UniGene
Hs.57690
Gene Chromosome Location
Chr.22: 26607982-26630672 on Build GRCh38
Transcripts
XM_006724140.3 NM_001887.3 XM_011529899.2

Gene Symbol
CRYBB1
Entrez Gene ID
1414
Gene Name
crystallin beta B1
Gene Aliases
CATCN3, CTRCT17
UniGene
Hs.37135
Gene Chromosome Location
Chr.22: 26599278-26618103 on Build GRCh38
Transcripts
XM_006724140.3 NM_001887.3 XM_011529899.2

SNP Detail

SNP ID Chromosome Location
rs932860964 Chr.22: 26617953-26617953 on Build GRCh38
rs767315034 Chr.22: 26617834-26617834 on Build GRCh38
rs755453252 Chr.22: 26618099-26618099 on Build GRCh38
rs923679465 Chr.22: 26617873-26617873 on Build GRCh38
rs1035884972 Chr.22: 26617872-26617872 on Build GRCh38
rs1036158876 Chr.22: 26618152-26618152 on Build GRCh38
rs766015917 Chr.22: 26618014-26618014 on Build GRCh38
rs796519448 Chr.22: 26617976-26617976 on Build GRCh38
rs970163791 Chr.22: 26617850-26617850 on Build GRCh38
rs967974150 Chr.22: 26618131-26618131 on Build GRCh38
rs780091439 Chr.22: 26618043-26618043 on Build GRCh38
rs909844257 Chr.22: 26617997-26617997 on Build GRCh38
rs558050790 Chr.22: 26617886-26617886 on Build GRCh38
rs747077193 Chr.22: 26618079-26618079 on Build GRCh38
rs201589516 Chr.22: 26617930-26617930 on Build GRCh38
rs886057327 Chr.22: 26618015-26618015 on Build GRCh38
rs1050012913 Chr.22: 26617988-26617988 on Build GRCh38
rs978866990 Chr.22: 26618012-26618012 on Build GRCh38
rs574199463 Chr.22: 26618110-26618110 on Build GRCh38
rs968960919 Chr.22: 26617977-26617977 on Build GRCh38
rs369210361 Chr.22: 26617796-26617796 on Build GRCh38
rs995914731 Chr.22: 26617805-26617805 on Build GRCh38
rs1022167144 Chr.22: 26618080-26618080 on Build GRCh38
rs977768278 Chr.22: 26617851-26617851 on Build GRCh38
rs931754716 Chr.22: 26618045-26618045 on Build GRCh38
rs573422267 Chr.22: 26617917-26617917 on Build GRCh38
rs1003652048 Chr.22: 26618044-26618044 on Build GRCh38
rs1001618786 Chr.22: 26617869-26617869 on Build GRCh38
rs987167823 Chr.22: 26618047-26618047 on Build GRCh38
rs750074449 Chr.22: 26617898-26617898 on Build GRCh38
rs922813336 Chr.22: 26617946-26617946 on Build GRCh38
rs999585749 Chr.22: 26618153-26618153 on Build GRCh38
rs1036700766 Chr.22: 26617770-26617770 on Build GRCh38
rs140796775 Chr.22: 26618012-26618012 on Build GRCh38
rs77926469 Chr.22: 26617820-26617820 on Build GRCh38
rs1012134678 Chr.22: 26617765-26617765 on Build GRCh38
rs1056675889 Chr.22: 26618046-26618046 on Build GRCh38
rs1037067727 Chr.22: 26618042-26618042 on Build GRCh38
rs9613229 Chr.22: 26618168-26618168 on Build GRCh38
rs753321812 Chr.22: 26618076-26618076 on Build GRCh38
rs1030690282 Chr.22: 26618222-26618222 on Build GRCh38
rs930568895 Chr.22: 26618173-26618173 on Build GRCh38
rs562125851 Chr.22: 26618050-26618050 on Build GRCh38

View Full Product Details