Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDLIM2
Entrez Gene ID
64236
Gene Name
PDZ and LIM domain 2
Gene Aliases
MYSTIQUE, SLIM
Chromosome Location
Chr.8: 22578741 - 22598025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021630.5 NP_067643.3 3 - 4 1034 104
NM_176871.3 NP_789847.1 2 - 3 376 104
NM_198042.3 NP_932159.1 2 - 3 376 104
GenBank mRNA AF289563.1 - 3 - 4 633 104
AK024498.1 - 2 - 3 319 104
AK027800.1 - 3 - 4 645 104
AK092968.1 - 3 - 4 1034 104
AY007729.1 - 2 - 3 400 104
AY070438.1 - 2 - 3 400 104
AY217349.1 - 2 - 3 361 104
BC021556.1 - 3 - 4 515 104
BC071774.1 - 3 - 4 645 104
DN997519.1 - 3 - 4 320 104

View Full Product Details