The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166247.1 | NP_001159719.1 | 3 - 4 | 838 | 135 |
NM_021956.4 | NP_068775.1 | 3 - 4 | 838 | 135 | |
NM_175768.3 | NP_786944.1 | 3 - 4 | 838 | 135 | |
XM_005266945.2 | XP_005267002.1 | 4 - 5 | 968 | 135 | |
XM_005266946.3 | XP_005267003.1 | 3 - 4 | 923 | 135 | |
XM_011535777.2 | XP_011534079.1 | 3 - 4 | 1005 | 135 | |
XM_011535778.2 | XP_011534080.1 | 3 - 4 | 1005 | 135 | |
XM_017010781.1 | XP_016866270.1 | 3 - 4 | 1005 | 135 | |
XM_017010782.1 | XP_016866271.1 | 3 - 4 | 1005 | 135 | |
GenBank mRNA | AJ252246.1 | - | 3 - 4 | 545 | 135 |
AJ301610.1 | - | 3 - 4 | 545 | 135 | |
AK289542.1 | - | 3 - 4 | 772 | 135 | |
AK291965.1 | - | 3 - 4 | 794 | 135 | |
AK303721.1 | - | 3 - 4 | 548 | 135 | |
BC037954.1 | - | 3 - 4 | 1030 | 135 | |
BC063814.1 | - | 3 - 4 | 748 | 135 | |
HM149335.1 | - | 3 - 4 | 545 | 135 | |
HM149336.1 | - | 3 - 4 | 545 | 135 | |
HM149337.1 | - | 3 - 4 | 545 | 135 | |
HM149338.1 | - | 3 - 4 | 545 | 135 | |
HM149339.1 | - | 3 - 4 | 545 | 135 | |
U16126.1 | - | 3 - 4 | 545 | 135 |