Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93894507-93894991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCLM
Entrez Gene ID
2730
Gene Name
glutamate-cysteine ligase modifier subunit
Gene Aliases
GLCLR
UniGene
Hs.315562
Gene Chromosome Location
Chr.1: 93885199-93909598 on Build GRCh38
Transcripts
NM_001308253.1 NM_002061.3 XM_011541261.2 XM_017001056.1 XM_017001057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3689891 c.635T>C p.L212S Chr.1: 93894634-93894634 on Build GRCh38
4686644 c.628C>T p.Q210* Chr.1: 93894641-93894641 on Build GRCh38
4010685 c.637A>G p.T213A Chr.1: 93894632-93894632 on Build GRCh38
1296809 c.568C>T p.L190F Chr.1: 93894701-93894701 on Build GRCh38
6881003 c.554G>A p.S185N Chr.1: 93894715-93894715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759970863 Chr.1: 93894627-93894627 on Build GRCh38
rs762713949 Chr.1: 93894677-93894677 on Build GRCh38
rs377180606 Chr.1: 93894752-93894752 on Build GRCh38
rs1046876763 Chr.1: 93894906-93894906 on Build GRCh38
rs769211 Chr.1: 93894551-93894551 on Build GRCh38
rs374378555 Chr.1: 93894842-93894843 on Build GRCh38
rs894398680 Chr.1: 93894964-93894964 on Build GRCh38
rs760114834 Chr.1: 93894653-93894654 on Build GRCh38
rs373013096 Chr.1: 93894772-93894772 on Build GRCh38
rs370374424 Chr.1: 93894775-93894775 on Build GRCh38
rs763622368 Chr.1: 93894693-93894693 on Build GRCh38
rs1025569150 Chr.1: 93894959-93894959 on Build GRCh38
rs557538608 Chr.1: 93894783-93894783 on Build GRCh38
rs750387132 Chr.1: 93894751-93894751 on Build GRCh38
rs1055744527 Chr.1: 93894841-93894841 on Build GRCh38
rs964270165 Chr.1: 93894600-93894600 on Build GRCh38
rs938552041 Chr.1: 93894832-93894832 on Build GRCh38
rs994228957 Chr.1: 93894958-93894958 on Build GRCh38
rs990376118 Chr.1: 93894605-93894605 on Build GRCh38
rs777846638 Chr.1: 93894766-93894766 on Build GRCh38
rs751772449 Chr.1: 93894825-93894825 on Build GRCh38
rs774931705 Chr.1: 93894669-93894669 on Build GRCh38
rs886921327 Chr.1: 93894914-93894914 on Build GRCh38
rs748408461 Chr.1: 93894563-93894563 on Build GRCh38
rs773668984 Chr.1: 93894574-93894574 on Build GRCh38
rs768552088 Chr.1: 93894775-93894776 on Build GRCh38
rs756986775 Chr.1: 93894717-93894717 on Build GRCh38
rs749438526 Chr.1: 93894956-93894956 on Build GRCh38
rs182660679 Chr.1: 93894782-93894782 on Build GRCh38
rs575801231 Chr.1: 93894815-93894815 on Build GRCh38
rs17880087 Chr.1: 93894642-93894642 on Build GRCh38
rs928303286 Chr.1: 93894828-93894829 on Build GRCh38
rs17879292 Chr.1: 93894929-93894929 on Build GRCh38
rs1006087131 Chr.1: 93894937-93894937 on Build GRCh38
rs187312951 Chr.1: 93894918-93894918 on Build GRCh38
rs758716981 Chr.1: 93894763-93894763 on Build GRCh38
rs553602888 Chr.1: 93894558-93894558 on Build GRCh38
rs771527985 Chr.1: 93894576-93894576 on Build GRCh38
rs545651979 Chr.1: 93894953-93894953 on Build GRCh38
rs540175616 Chr.1: 93894961-93894961 on Build GRCh38
rs748210352 Chr.1: 93894761-93894761 on Build GRCh38
rs898540078 Chr.1: 93894957-93894957 on Build GRCh38
rs764757939 Chr.1: 93894646-93894646 on Build GRCh38
rs929412050 Chr.1: 93894878-93894878 on Build GRCh38
rs751435161 Chr.1: 93894705-93894705 on Build GRCh38
rs781697717 Chr.1: 93894613-93894613 on Build GRCh38
rs756430538 Chr.1: 93894932-93894932 on Build GRCh38
rs777135699 Chr.1: 93894625-93894625 on Build GRCh38
rs2234730 Chr.1: 93894760-93894760 on Build GRCh38
rs767358231 Chr.1: 93894720-93894720 on Build GRCh38
rs923374222 Chr.1: 93894803-93894803 on Build GRCh38
rs910814612 Chr.1: 93894611-93894611 on Build GRCh38
rs17881946 Chr.1: 93894535-93894535 on Build GRCh38
rs546253959 Chr.1: 93894860-93894860 on Build GRCh38
rs772164973 Chr.1: 93894564-93894564 on Build GRCh38
rs772523300 Chr.1: 93894757-93894759 on Build GRCh38
rs77237202 Chr.1: 93894538-93894538 on Build GRCh38

View Full Product Details