Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93993140-93993643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA4
Entrez Gene ID
24
Gene Name
ATP binding cassette subfamily A member 4
Gene Aliases
ABC10, ABCR, ARMD2, CORD3, FFM, RMP, RP19, STGD, STGD1
UniGene
Hs.416707
Gene Chromosome Location
Chr.1: 93992837-94121149 on Build GRCh38
Transcripts
NM_000350.2

SNP Detail

SNP ID Chromosome Location
rs750565255 Chr.1: 93993602-93993602 on Build GRCh38
rs767072316 Chr.1: 93993193-93993193 on Build GRCh38
rs370860160 Chr.1: 93993267-93993267 on Build GRCh38
rs986430094 Chr.1: 93993595-93993595 on Build GRCh38
rs755828662 Chr.1: 93993203-93993203 on Build GRCh38
rs1019667491 Chr.1: 93993461-93993461 on Build GRCh38
rs955079945 Chr.1: 93993221-93993221 on Build GRCh38
rs866960838 Chr.1: 93993270-93993270 on Build GRCh38
rs765812753 Chr.1: 93993283-93993283 on Build GRCh38
rs145634012 Chr.1: 93993227-93993227 on Build GRCh38
rs767876749 Chr.1: 93993268-93993268 on Build GRCh38
rs895403241 Chr.1: 93993477-93993477 on Build GRCh38
rs1012890108 Chr.1: 93993480-93993480 on Build GRCh38
rs769173189 Chr.1: 93993260-93993260 on Build GRCh38
rs754777067 Chr.1: 93993220-93993220 on Build GRCh38
rs563707924 Chr.1: 93993564-93993564 on Build GRCh38
rs560969749 Chr.1: 93993403-93993403 on Build GRCh38
rs926313696 Chr.1: 93993607-93993607 on Build GRCh38
rs541209823 Chr.1: 93993392-93993392 on Build GRCh38
rs766623453 Chr.1: 93993176-93993176 on Build GRCh38
rs367853124 Chr.1: 93993275-93993275 on Build GRCh38
rs369306889 Chr.1: 93993222-93993222 on Build GRCh38
rs201835773 Chr.1: 93993185-93993185 on Build GRCh38
rs1025380942 Chr.1: 93993575-93993575 on Build GRCh38
rs754576845 Chr.1: 93993289-93993289 on Build GRCh38
rs919146588 Chr.1: 93993162-93993162 on Build GRCh38
rs937652642 Chr.1: 93993608-93993608 on Build GRCh38
rs566725549 Chr.1: 93993376-93993377 on Build GRCh38
rs377008865 Chr.1: 93993226-93993226 on Build GRCh38
rs749875918 Chr.1: 93993198-93993198 on Build GRCh38
rs201160433 Chr.1: 93993182-93993182 on Build GRCh38
rs577038211 Chr.1: 93993335-93993335 on Build GRCh38
rs966279875 Chr.1: 93993475-93993475 on Build GRCh38
rs75999881 Chr.1: 93993327-93993327 on Build GRCh38
rs375731558 Chr.1: 93993269-93993269 on Build GRCh38
rs779151391 Chr.1: 93993550-93993550 on Build GRCh38
rs775865861 Chr.1: 93993243-93993243 on Build GRCh38
rs753523367 Chr.1: 93993286-93993286 on Build GRCh38
rs889773161 Chr.1: 93993424-93993424 on Build GRCh38
rs200205293 Chr.1: 93993277-93993277 on Build GRCh38
rs762183450 Chr.1: 93993188-93993188 on Build GRCh38
rs748141123 Chr.1: 93993224-93993224 on Build GRCh38
rs546533237 Chr.1: 93993613-93993613 on Build GRCh38
rs945851538 Chr.1: 93993348-93993348 on Build GRCh38
rs200401067 Chr.1: 93993214-93993214 on Build GRCh38
rs575090193 Chr.1: 93993238-93993238 on Build GRCh38
rs1056835452 Chr.1: 93993467-93993468 on Build GRCh38
rs17110718 Chr.1: 93993444-93993444 on Build GRCh38
rs368493917 Chr.1: 93993303-93993303 on Build GRCh38
rs376708072 Chr.1: 93993261-93993261 on Build GRCh38
rs902598948 Chr.1: 93993384-93993384 on Build GRCh38
rs574870927 Chr.1: 93993464-93993464 on Build GRCh38
rs372398535 Chr.1: 93993225-93993225 on Build GRCh38
rs200946608 Chr.1: 93993211-93993211 on Build GRCh38
rs962505043 Chr.1: 93993183-93993183 on Build GRCh38
rs78488259 Chr.1: 93993520-93993520 on Build GRCh38
rs749454214 Chr.1: 93993259-93993259 on Build GRCh38
rs951998114 Chr.1: 93993588-93993588 on Build GRCh38

View Full Product Details