Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94007651-94008181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA4
Entrez Gene ID
24
Gene Name
ATP binding cassette subfamily A member 4
Gene Aliases
ABC10, ABCR, ARMD2, CORD3, FFM, RMP, RP19, STGD, STGD1
UniGene
Hs.416707
Gene Chromosome Location
Chr.1: 93992837-94121149 on Build GRCh38
Transcripts
NM_000350.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246290 c.5907C>A p.G1969G Chr.1: 94007732-94007732 on Build GRCh38
7353496 c.718C>T p.L240F Chr.1: 94007731-94007731 on Build GRCh38
3790355 c.5937A>C p.T1979T Chr.1: 94007702-94007702 on Build GRCh38
7122388 c.747A>C p.T249T Chr.1: 94007702-94007702 on Build GRCh38
6262109 c.5961G>T p.G1987G Chr.1: 94007678-94007678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61750640 Chr.1: 94007675-94007678 on Build GRCh38
rs745851261 Chr.1: 94007733-94007733 on Build GRCh38
rs886046559 Chr.1: 94007686-94007686 on Build GRCh38
rs61750639 Chr.1: 94007710-94007710 on Build GRCh38
rs116198846 Chr.1: 94008148-94008148 on Build GRCh38
rs894553905 Chr.1: 94007976-94007976 on Build GRCh38
rs751410605 Chr.1: 94007762-94007762 on Build GRCh38
rs3789385 Chr.1: 94008001-94008001 on Build GRCh38
rs3789384 Chr.1: 94007923-94007923 on Build GRCh38
rs952794296 Chr.1: 94008131-94008131 on Build GRCh38
rs533467925 Chr.1: 94007770-94007770 on Build GRCh38
rs61753034 Chr.1: 94007727-94007727 on Build GRCh38
rs1041683476 Chr.1: 94007874-94007874 on Build GRCh38
rs756012892 Chr.1: 94007758-94007758 on Build GRCh38
rs940312393 Chr.1: 94007866-94007866 on Build GRCh38
rs761937846 Chr.1: 94007759-94007759 on Build GRCh38
rs934236448 Chr.1: 94007918-94007918 on Build GRCh38
rs35156284 Chr.1: 94007680-94007681 on Build GRCh38
rs764881591 Chr.1: 94007690-94007690 on Build GRCh38
rs538742590 Chr.1: 94008057-94008057 on Build GRCh38
rs1030634915 Chr.1: 94008120-94008120 on Build GRCh38
rs558090925 Chr.1: 94008009-94008011 on Build GRCh38
rs531446153 Chr.1: 94007944-94007944 on Build GRCh38
rs982210305 Chr.1: 94007806-94007806 on Build GRCh38
rs915413568 Chr.1: 94007855-94007855 on Build GRCh38
rs202088666 Chr.1: 94007717-94007717 on Build GRCh38
rs941429154 Chr.1: 94007956-94007956 on Build GRCh38
rs956470370 Chr.1: 94007847-94007847 on Build GRCh38
rs989158765 Chr.1: 94007849-94007849 on Build GRCh38
rs1010525543 Chr.1: 94008151-94008151 on Build GRCh38
rs886044756 Chr.1: 94007730-94007730 on Build GRCh38
rs766811018 Chr.1: 94007673-94007673 on Build GRCh38
rs770010375 Chr.1: 94007813-94007813 on Build GRCh38
rs758129527 Chr.1: 94007699-94007699 on Build GRCh38
rs116391773 Chr.1: 94007887-94007887 on Build GRCh38
rs142123094 Chr.1: 94007698-94007698 on Build GRCh38
rs747180153 Chr.1: 94007789-94007789 on Build GRCh38
rs28938473 Chr.1: 94007731-94007731 on Build GRCh38
rs376767237 Chr.1: 94007711-94007711 on Build GRCh38
rs149063807 Chr.1: 94007992-94007995 on Build GRCh38
rs556341538 Chr.1: 94007875-94007875 on Build GRCh38
rs61753037 Chr.1: 94007703-94007703 on Build GRCh38
rs201545948 Chr.1: 94007684-94007684 on Build GRCh38
rs1003435093 Chr.1: 94008115-94008115 on Build GRCh38
rs752147871 Chr.1: 94007697-94007697 on Build GRCh38
rs773339084 Chr.1: 94007785-94007785 on Build GRCh38
rs551430410 Chr.1: 94007841-94007841 on Build GRCh38
rs200579529 Chr.1: 94007720-94007720 on Build GRCh38
rs61753035 Chr.1: 94007725-94007725 on Build GRCh38
rs61751389 Chr.1: 94007722-94007722 on Build GRCh38
rs61753036 Chr.1: 94007716-94007716 on Build GRCh38
rs980881735 Chr.1: 94007675-94007675 on Build GRCh38
rs975600791 Chr.1: 94007882-94007882 on Build GRCh38
rs145297440 Chr.1: 94008089-94008089 on Build GRCh38
rs765957871 Chr.1: 94007957-94007957 on Build GRCh38
rs1030602448 Chr.1: 94007832-94007832 on Build GRCh38
rs368609612 Chr.1: 94007846-94007846 on Build GRCh38
rs3789386 Chr.1: 94008049-94008049 on Build GRCh38
rs112375682 Chr.1: 94007966-94007966 on Build GRCh38
rs61748516 Chr.1: 94007687-94007688 on Build GRCh38
rs1019219577 Chr.1: 94007865-94007865 on Build GRCh38
rs867079318 Chr.1: 94007709-94007709 on Build GRCh38
rs760098992 Chr.1: 94007682-94007682 on Build GRCh38
rs1024604313 Chr.1: 94008016-94008016 on Build GRCh38
rs560127680 Chr.1: 94007842-94007842 on Build GRCh38
rs750031062 Chr.1: 94007880-94007880 on Build GRCh38
rs777300047 Chr.1: 94007680-94007680 on Build GRCh38
rs1049484456 Chr.1: 94007912-94007912 on Build GRCh38
rs1022034794 Chr.1: 94008155-94008155 on Build GRCh38
rs952702814 Chr.1: 94007730-94007730 on Build GRCh38
rs763801033 Chr.1: 94008125-94008125 on Build GRCh38
rs753308390 Chr.1: 94007970-94007970 on Build GRCh38
rs369431053 Chr.1: 94007777-94007777 on Build GRCh38
rs574212346 Chr.1: 94007930-94007930 on Build GRCh38

View Full Product Details