The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022072.3 | NP_071355.1 | 3 - 4 | 585 | 84 |
GenBank mRNA | AF169972.1 | - | 3 - 4 | 594 | 84 |
AK026262.1 | - | 3 - 4 | 577 | 84 | |
BC020602.1 | - | 3 - 4 | 585 | 84 | |
BC057238.1 | - | 3 - 4 | 589 | 84 |