Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLSPN
Entrez Gene ID
63967
Gene Name
claspin
Gene Aliases
-
Chromosome Location
Chr.1: 35720465 - 35769985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011541939.2 XP_011540241.1 24 - 25 4007 61
GenBank mRNA AF297866.1 - 24 - 25 3990 61

View Full Product Details