The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022437.2 | NP_071882.1 | 6 - 7 | 1051 | 63 |
XM_005264483.3 | XP_005264540.1 | 6 - 7 | 1166 | 63 | |
XM_011533029.2 | XP_011531331.1 | 6 - 7 | 1618 | 63 | |
XM_011533030.2 | XP_011531332.1 | 6 - 7 | 1463 | 63 | |
GenBank mRNA | AF320294.1 | - | 6 - 7 | 961 | 63 |
AF324494.1 | - | 6 - 7 | 1051 | 63 | |
BC113657.1 | - | 6 - 7 | 997 | 63 | |
BC143564.1 | - | 6 - 7 | 997 | 63 |