Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38115447-38115947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
UniGene
Hs.170479
Gene Chromosome Location
Chr.22: 38111495-38192099 on Build GRCh38
Transcripts
NM_001004426.1 NM_001199562.1 NM_003560.2 XM_005261764.2 XM_005261765.1 XM_005261766.1 XM_006724332.3 XM_011530422.1 XM_011530423.1 XM_011530424.1 XM_011530425.1 XM_017028981.1 XM_017028982.1 XM_017028983.1 XM_017028984.1 XM_017028985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4475535 c.1997C>T p.T666I Chr.22: 38115564-38115564 on Build GRCh38
1220860 c.1904G>A p.R635Q Chr.22: 38115657-38115657 on Build GRCh38
7159612 c.1942G>A p.G648R Chr.22: 38115619-38115619 on Build GRCh38
6860189 c.1932C>T p.F644F Chr.22: 38115629-38115629 on Build GRCh38
7384179 c.2030G>A p.R677H Chr.22: 38115531-38115531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779376537 Chr.22: 38115605-38115605 on Build GRCh38
rs928876814 Chr.22: 38115521-38115521 on Build GRCh38
rs1023196935 Chr.22: 38115781-38115781 on Build GRCh38
rs587784340 Chr.22: 38115588-38115588 on Build GRCh38
rs908659767 Chr.22: 38115595-38115595 on Build GRCh38
rs754204295 Chr.22: 38115604-38115604 on Build GRCh38
rs977648892 Chr.22: 38115917-38115917 on Build GRCh38
rs768064585 Chr.22: 38115757-38115757 on Build GRCh38
rs755814729 Chr.22: 38115906-38115907 on Build GRCh38
rs776500207 Chr.22: 38115901-38115901 on Build GRCh38
rs777074144 Chr.22: 38115837-38115837 on Build GRCh38
rs762340313 Chr.22: 38115846-38115846 on Build GRCh38
rs780784951 Chr.22: 38115627-38115627 on Build GRCh38
rs1034806198 Chr.22: 38115650-38115650 on Build GRCh38
rs150572286 Chr.22: 38115578-38115578 on Build GRCh38
rs1041927697 Chr.22: 38115799-38115799 on Build GRCh38
rs932664861 Chr.22: 38115867-38115867 on Build GRCh38
rs370975770 Chr.22: 38115666-38115666 on Build GRCh38
rs922289885 Chr.22: 38115916-38115916 on Build GRCh38
rs952875012 Chr.22: 38115783-38115783 on Build GRCh38
rs112240186 Chr.22: 38115913-38115913 on Build GRCh38
rs570033641 Chr.22: 38115689-38115689 on Build GRCh38
rs754114238 Chr.22: 38115700-38115700 on Build GRCh38
rs765557069 Chr.22: 38115875-38115875 on Build GRCh38
rs900909906 Chr.22: 38115694-38115694 on Build GRCh38
rs1018063644 Chr.22: 38115888-38115888 on Build GRCh38
rs370731849 Chr.22: 38115822-38115822 on Build GRCh38
rs374942810 Chr.22: 38115517-38115517 on Build GRCh38
rs754415689 Chr.22: 38115621-38115621 on Build GRCh38
rs1025497590 Chr.22: 38115682-38115682 on Build GRCh38
rs764292621 Chr.22: 38115699-38115699 on Build GRCh38
rs910928544 Chr.22: 38115737-38115737 on Build GRCh38
rs765630635 Chr.22: 38115703-38115703 on Build GRCh38
rs1041011926 Chr.22: 38115632-38115632 on Build GRCh38
rs767459234 Chr.22: 38115677-38115677 on Build GRCh38
rs539530139 Chr.22: 38115905-38115905 on Build GRCh38
rs976385779 Chr.22: 38115763-38115763 on Build GRCh38
rs1005791518 Chr.22: 38115880-38115880 on Build GRCh38
rs571716373 Chr.22: 38115838-38115838 on Build GRCh38
rs183446770 Chr.22: 38115563-38115563 on Build GRCh38
rs772024534 Chr.22: 38115676-38115678 on Build GRCh38
rs767689496 Chr.22: 38115579-38115579 on Build GRCh38
rs377488420 Chr.22: 38115659-38115659 on Build GRCh38
rs746514704 Chr.22: 38115532-38115532 on Build GRCh38
rs922606379 Chr.22: 38115856-38115856 on Build GRCh38
rs746460352 Chr.22: 38115742-38115742 on Build GRCh38
rs999011583 Chr.22: 38115726-38115726 on Build GRCh38
rs904692346 Chr.22: 38115810-38115810 on Build GRCh38
rs200280060 Chr.22: 38115662-38115662 on Build GRCh38
rs1021782317 Chr.22: 38115701-38115701 on Build GRCh38
rs977256373 Chr.22: 38115855-38115855 on Build GRCh38
rs201657455 Chr.22: 38115663-38115663 on Build GRCh38
rs121908683 Chr.22: 38115667-38115667 on Build GRCh38
rs369038599 Chr.22: 38115531-38115531 on Build GRCh38
rs533490649 Chr.22: 38115635-38115635 on Build GRCh38
rs387906863 Chr.22: 38115657-38115657 on Build GRCh38
rs730882214 Chr.22: 38115650-38115650 on Build GRCh38
rs368948611 Chr.22: 38115709-38115709 on Build GRCh38
rs902970540 Chr.22: 38115720-38115720 on Build GRCh38
rs1055915360 Chr.22: 38115823-38115823 on Build GRCh38
rs1000303487 Chr.22: 38115649-38115649 on Build GRCh38
rs376610322 Chr.22: 38115572-38115572 on Build GRCh38
rs1035327284 Chr.22: 38115727-38115727 on Build GRCh38
rs751614625 Chr.22: 38115569-38115569 on Build GRCh38
rs769498964 Chr.22: 38115648-38115648 on Build GRCh38
rs764396606 Chr.22: 38115590-38115590 on Build GRCh38
rs753021482 Chr.22: 38115491-38115491 on Build GRCh38
rs375645986 Chr.22: 38115690-38115690 on Build GRCh38
rs991708493 Chr.22: 38115556-38115556 on Build GRCh38
rs1025530217 Chr.22: 38115859-38115859 on Build GRCh38
rs746432450 Chr.22: 38115616-38115616 on Build GRCh38
rs774856860 Chr.22: 38115478-38115478 on Build GRCh38
rs12627857 Chr.22: 38115577-38115577 on Build GRCh38
rs773162246 Chr.22: 38115558-38115558 on Build GRCh38
rs202193870 Chr.22: 38115518-38115518 on Build GRCh38
rs75406900 Chr.22: 38115474-38115474 on Build GRCh38
rs1050322812 Chr.22: 38115907-38115907 on Build GRCh38
rs926187509 Chr.22: 38115919-38115919 on Build GRCh38
rs565073323 Chr.22: 38115832-38115833 on Build GRCh38
rs772904835 Chr.22: 38115676-38115676 on Build GRCh38
rs553624681 Chr.22: 38115780-38115780 on Build GRCh38
rs757592075 Chr.22: 38115702-38115702 on Build GRCh38
rs564126033 Chr.22: 38115471-38115471 on Build GRCh38
rs794729212 Chr.22: 38115619-38115619 on Build GRCh38
rs776124801 Chr.22: 38115536-38115536 on Build GRCh38
rs959711603 Chr.22: 38115741-38115741 on Build GRCh38
rs547175863 Chr.22: 38115582-38115582 on Build GRCh38
rs749757274 Chr.22: 38115527-38115527 on Build GRCh38
rs561645994 Chr.22: 38115533-38115533 on Build GRCh38
rs587784341 Chr.22: 38115583-38115583 on Build GRCh38
rs1048444597 Chr.22: 38115587-38115587 on Build GRCh38
rs535260587 Chr.22: 38115775-38115775 on Build GRCh38
rs1021400208 Chr.22: 38115469-38115469 on Build GRCh38
rs201727354 Chr.22: 38115584-38115584 on Build GRCh38
rs369680655 Chr.22: 38115546-38115546 on Build GRCh38
rs1014278404 Chr.22: 38115833-38115833 on Build GRCh38
rs769434263 Chr.22: 38115551-38115551 on Build GRCh38
rs760482527 Chr.22: 38115684-38115684 on Build GRCh38
rs567944607 Chr.22: 38115711-38115711 on Build GRCh38
rs557521572 Chr.22: 38115922-38115922 on Build GRCh38
rs762928103 Chr.22: 38115562-38115562 on Build GRCh38

View Full Product Details