Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS33A
Entrez Gene ID
65082
Gene Name
VPS33A, CORVET/HOPS core subunit
Gene Aliases
-
Chromosome Location
Chr.12: 122231547 - 122266521 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022916.4 NP_075067.2 11 - 12 1553 69
GenBank mRNA AF439857.1 - 11 - 12 1464 69
AK026048.1 - 5 - 6 674 69
AK026840.1 - 11 - 12 1463 69
AK315545.1 - 11 - 12 1491 69
BC016617.1 - 11 - 12 1461 69

View Full Product Details