The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293285.1 | NP_001280214.1 | 10 - 11 | 1913 | 62 |
NM_001293286.1 | NP_001280215.1 | 8 - 9 | 1766 | 62 | |
NM_023071.3 | NP_075559.2 | 9 - 10 | 1829 | 62 | |
XM_011538690.2 | XP_011536992.1 | 10 - 11 | 1301 | 62 | |
XM_011538691.2 | XP_011536993.1 | 8 - 9 | 1196 | 62 | |
XM_017019840.1 | XP_016875329.1 | 9 - 10 | 1269 | 62 | |
GenBank mRNA | AB075515.1 | - | 7 - 8 | 857 | 62 |
AK023179.1 | - | 5 - 6 | 738 | 62 | |
AK023202.1 | - | 9 - 10 | 1228 | 62 | |
AK024678.1 | - | 1 - 2 | 125 | 62 | |
AK292334.1 | - | 8 - 9 | 1166 | 62 | |
AL833614.1 | - | 10 - 11 | 1354 | 62 | |
BC048299.1 | - | 8 - 9 | 1196 | 62 |