The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286655.1 | NP_001273584.1 | 3 - 4 | 842 | 119 |
NM_001286656.1 | NP_001273585.1 | 3 - 4 | 426 | 119 | |
NM_023932.3 | NP_076421.2 | 3 - 4 | 426 | 119 | |
NM_206539.2 | NP_996262.1 | 3 - 4 | 842 | 119 | |
XM_005249308.4 | XP_005249365.1 | 2 - 3 | 1634 | 119 | |
XM_011514823.2 | XP_011513125.1 | 2 - 3 | 1088 | 119 | |
GenBank mRNA | AK055380.1 | - | 3 - 4 | 842 | 119 |
AY358126.1 | - | 3 - 4 | 292 | 119 | |
BC000230.1 | - | 3 - 4 | 258 | 119 | |
BC006425.1 | - | 3 - 4 | 258 | 119 | |
BC110320.1 | - | 3 - 4 | 394 | 119 | |
BI916137.1 | - | 3 - 4 | 350 | 119 | |
BM811564.1 | - | - | 391 | 119 | |
DA332624.1 | - | 3 - 4 | 293 | 119 |