Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40359118-40359528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSL
Entrez Gene ID
158
Gene Name
adenylosuccinate lyase
Gene Aliases
AMPS, ASASE, ASL
UniGene
Hs.75527
Gene Chromosome Location
Chr.22: 40346500-40387408 on Build GRCh38
Transcripts
NM_000026.3 NM_001123378.2 NM_001317923.1 NR_134256.1 XM_011529977.2 XM_011529979.1 XM_011529980.2 XM_017028636.1 XM_017028637.1 XM_017028638.1 XM_017028639.1 XM_017028640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034431 c.701G>A p.R234K Chr.22: 40359306-40359306 on Build GRCh38
7128621 c.711C>G p.D237E Chr.22: 40359274-40359274 on Build GRCh38
1484257 c.663G>A p.Q221Q Chr.22: 40359268-40359268 on Build GRCh38
1308194 c.669C>G p.D223E Chr.22: 40359274-40359274 on Build GRCh38
1632616 c.668A>G p.D223G Chr.22: 40359273-40359273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746657973 Chr.22: 40359235-40359235 on Build GRCh38
rs887933781 Chr.22: 40359149-40359149 on Build GRCh38
rs1021146802 Chr.22: 40359191-40359191 on Build GRCh38
rs759768270 Chr.22: 40359344-40359344 on Build GRCh38
rs376028329 Chr.22: 40359424-40359424 on Build GRCh38
rs775951751 Chr.22: 40359357-40359357 on Build GRCh38
rs8192459 Chr.22: 40359164-40359164 on Build GRCh38
rs1002640319 Chr.22: 40359240-40359240 on Build GRCh38
rs114141894 Chr.22: 40359147-40359147 on Build GRCh38
rs71321193 Chr.22: 40359350-40359350 on Build GRCh38
rs375709749 Chr.22: 40359335-40359335 on Build GRCh38
rs929574573 Chr.22: 40359423-40359423 on Build GRCh38
rs772989069 Chr.22: 40359310-40359310 on Build GRCh38
rs771546241 Chr.22: 40359146-40359146 on Build GRCh38
rs538222513 Chr.22: 40359406-40359406 on Build GRCh38
rs780906231 Chr.22: 40359249-40359249 on Build GRCh38
rs372676656 Chr.22: 40359322-40359322 on Build GRCh38
rs957197920 Chr.22: 40359342-40359342 on Build GRCh38
rs748105288 Chr.22: 40359306-40359306 on Build GRCh38
rs546878201 Chr.22: 40359307-40359307 on Build GRCh38
rs1048001718 Chr.22: 40359447-40359447 on Build GRCh38
rs56719087 Chr.22: 40359497-40359497 on Build GRCh38
rs768010664 Chr.22: 40359356-40359356 on Build GRCh38
rs748862331 Chr.22: 40359174-40359174 on Build GRCh38
rs899842356 Chr.22: 40359145-40359145 on Build GRCh38
rs374477575 Chr.22: 40359476-40359476 on Build GRCh38
rs371075097 Chr.22: 40359217-40359217 on Build GRCh38
rs899544918 Chr.22: 40359176-40359177 on Build GRCh38
rs1026848957 Chr.22: 40359403-40359403 on Build GRCh38
rs991155327 Chr.22: 40359345-40359345 on Build GRCh38
rs983001406 Chr.22: 40359496-40359496 on Build GRCh38
rs570943329 Chr.22: 40359397-40359397 on Build GRCh38
rs754771022 Chr.22: 40359247-40359247 on Build GRCh38
rs119450945 Chr.22: 40359279-40359279 on Build GRCh38
rs760352117 Chr.22: 40359390-40359390 on Build GRCh38
rs73885691 Chr.22: 40359419-40359419 on Build GRCh38
rs1040958171 Chr.22: 40359497-40359497 on Build GRCh38
rs770899805 Chr.22: 40359325-40359325 on Build GRCh38
rs774582275 Chr.22: 40359451-40359451 on Build GRCh38
rs779732422 Chr.22: 40359227-40359227 on Build GRCh38
rs1035728500 Chr.22: 40359241-40359241 on Build GRCh38
rs952657860 Chr.22: 40359404-40359404 on Build GRCh38
rs916006424 Chr.22: 40359383-40359383 on Build GRCh38
rs928611155 Chr.22: 40359423-40359424 on Build GRCh38
rs138661558 Chr.22: 40359218-40359218 on Build GRCh38
rs534015761 Chr.22: 40359166-40359166 on Build GRCh38
rs377555022 Chr.22: 40359212-40359212 on Build GRCh38
rs201172209 Chr.22: 40359476-40359476 on Build GRCh38

View Full Product Details