Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf49
Entrez Gene ID
79074
Gene Name
chromosome 2 open reading frame 49
Gene Aliases
asw
Chromosome Location
Chr.2: 105337257 - 105359940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286537.1 NP_001273466.1 4 - 5 743 128
NM_024093.2 NP_076998.1 3 - 4 869 128
XM_017004892.1 XP_016860381.1 4 - 5 845 128
GenBank mRNA AK127661.1 - 3 - 4 674 128
AK304268.1 - 4 - 5 743 128
AK307891.1 - 4 - 5 569 128
BC001310.1 - 3 - 4 672 128

View Full Product Details